Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141339680-141339923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_032009.2

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_032009.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4712406 c.796G>A p.A266T Chr.5: 141339767-141339767 on Build GRCh38
4386264 c.748A>C p.S250R Chr.5: 141339719-141339719 on Build GRCh38
1062975 c.743G>A p.R248H Chr.5: 141339714-141339714 on Build GRCh38
1287018 c.879G>A p.E293E Chr.5: 141339850-141339850 on Build GRCh38
6846705 c.852C>G p.S284R Chr.5: 141339823-141339823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748786219 Chr.5: 141339851-141339852 on Build GRCh38
rs752711661 Chr.5: 141339869-141339869 on Build GRCh38
rs769034523 Chr.5: 141339800-141339800 on Build GRCh38
rs764027282 Chr.5: 141339876-141339876 on Build GRCh38
rs150706052 Chr.5: 141339716-141339716 on Build GRCh38
rs750215738 Chr.5: 141339768-141339768 on Build GRCh38
rs753811865 Chr.5: 141339810-141339810 on Build GRCh38
rs201553091 Chr.5: 141339750-141339750 on Build GRCh38
rs1026704695 Chr.5: 141339745-141339745 on Build GRCh38
rs760007319 Chr.5: 141339713-141339713 on Build GRCh38
rs145851665 Chr.5: 141339749-141339749 on Build GRCh38
rs747337988 Chr.5: 141339796-141339796 on Build GRCh38
rs759098844 Chr.5: 141339848-141339848 on Build GRCh38
rs141252229 Chr.5: 141339874-141339874 on Build GRCh38
rs753788079 Chr.5: 141339881-141339882 on Build GRCh38
rs143562227 Chr.5: 141339720-141339720 on Build GRCh38
rs368191708 Chr.5: 141339863-141339863 on Build GRCh38
rs762403810 Chr.5: 141339721-141339721 on Build GRCh38
rs867776152 Chr.5: 141339765-141339765 on Build GRCh38
rs761046052 Chr.5: 141339717-141339717 on Build GRCh38
rs200504537 Chr.5: 141339805-141339805 on Build GRCh38
rs754079141 Chr.5: 141339885-141339885 on Build GRCh38
rs760597232 Chr.5: 141339843-141339844 on Build GRCh38
rs1030624939 Chr.5: 141339782-141339782 on Build GRCh38
rs778896419 Chr.5: 141339776-141339776 on Build GRCh38
rs781028162 Chr.5: 141339736-141339736 on Build GRCh38
rs751484386 Chr.5: 141339733-141339733 on Build GRCh38
rs765909769 Chr.5: 141339827-141339827 on Build GRCh38
rs752482688 Chr.5: 141339739-141339739 on Build GRCh38
rs138877576 Chr.5: 141339764-141339764 on Build GRCh38
rs762635972 Chr.5: 141339825-141339825 on Build GRCh38
rs754938154 Chr.5: 141339735-141339735 on Build GRCh38
rs772509350 Chr.5: 141339786-141339786 on Build GRCh38
rs767349626 Chr.5: 141339706-141339707 on Build GRCh38
rs139043857 Chr.5: 141339791-141339791 on Build GRCh38
rs774870637 Chr.5: 141339706-141339706 on Build GRCh38
rs894686520 Chr.5: 141339707-141339707 on Build GRCh38
rs914988246 Chr.5: 141339841-141339841 on Build GRCh38
rs200162193 Chr.5: 141339813-141339813 on Build GRCh38
rs746379195 Chr.5: 141339784-141339784 on Build GRCh38
rs765859037 Chr.5: 141339727-141339727 on Build GRCh38
rs773791186 Chr.5: 141339829-141339829 on Build GRCh38
rs757484303 Chr.5: 141339889-141339889 on Build GRCh38

View Full Product Details