Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73631999-73632228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOZ1
Entrez Gene ID
58529
Gene Name
myozenin 1
Gene Aliases
CS-2, FATZ, MYOZ
UniGene
Hs.238756
Gene Chromosome Location
Chr.10: 73631612-73641757 on Build GRCh38
Transcripts
NM_021245.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4144852 c.720C>A p.F240L Chr.10: 73632110-73632110 on Build GRCh38
3440460 c.688G>A p.G230R Chr.10: 73632142-73632142 on Build GRCh38
5816254 c.764T>C p.L255P Chr.10: 73632066-73632066 on Build GRCh38
6842430 c.764T>A p.L255Q Chr.10: 73632066-73632066 on Build GRCh38
4015768 c.710G>A p.R237H Chr.10: 73632120-73632120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007129007 Chr.10: 73632096-73632096 on Build GRCh38
rs778393682 Chr.10: 73632028-73632030 on Build GRCh38
rs759629947 Chr.10: 73632024-73632024 on Build GRCh38
rs199706644 Chr.10: 73632063-73632063 on Build GRCh38
rs779736355 Chr.10: 73632071-73632071 on Build GRCh38
rs774593816 Chr.10: 73632022-73632022 on Build GRCh38
rs765804601 Chr.10: 73632175-73632175 on Build GRCh38
rs923399819 Chr.10: 73632177-73632177 on Build GRCh38
rs986757941 Chr.10: 73632196-73632196 on Build GRCh38
rs764489925 Chr.10: 73632046-73632046 on Build GRCh38
rs772257893 Chr.10: 73632137-73632137 on Build GRCh38
rs868140755 Chr.10: 73632072-73632072 on Build GRCh38
rs773215819 Chr.10: 73632120-73632120 on Build GRCh38
rs910824704 Chr.10: 73632200-73632200 on Build GRCh38
rs779337765 Chr.10: 73632055-73632055 on Build GRCh38
rs775615695 Chr.10: 73632142-73632142 on Build GRCh38
rs906601362 Chr.10: 73632031-73632031 on Build GRCh38
rs752951846 Chr.10: 73632027-73632027 on Build GRCh38
rs138613016 Chr.10: 73632115-73632115 on Build GRCh38
rs747847419 Chr.10: 73632109-73632109 on Build GRCh38
rs746647132 Chr.10: 73632077-73632077 on Build GRCh38
rs764175440 Chr.10: 73632156-73632156 on Build GRCh38
rs150166405 Chr.10: 73632088-73632088 on Build GRCh38
rs372228357 Chr.10: 73632052-73632052 on Build GRCh38
rs149293567 Chr.10: 73632121-73632121 on Build GRCh38
rs147643592 Chr.10: 73632021-73632021 on Build GRCh38
rs762210190 Chr.10: 73632089-73632089 on Build GRCh38
rs375583471 Chr.10: 73632017-73632017 on Build GRCh38
rs767855451 Chr.10: 73632025-73632025 on Build GRCh38
rs747844516 Chr.10: 73632101-73632101 on Build GRCh38
rs41280400 Chr.10: 73632065-73632065 on Build GRCh38
rs771183781 Chr.10: 73632178-73632182 on Build GRCh38
rs750920977 Chr.10: 73632192-73632192 on Build GRCh38
rs1018147208 Chr.10: 73632119-73632119 on Build GRCh38
rs376703108 Chr.10: 73632159-73632159 on Build GRCh38
rs781069032 Chr.10: 73632101-73632101 on Build GRCh38
rs888378503 Chr.10: 73632091-73632091 on Build GRCh38
rs760989673 Chr.10: 73632153-73632153 on Build GRCh38
rs757949747 Chr.10: 73632195-73632195 on Build GRCh38
rs760994203 Chr.10: 73632045-73632045 on Build GRCh38
rs777071432 Chr.10: 73632158-73632158 on Build GRCh38

View Full Product Details