Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52381286-52381551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT84
Entrez Gene ID
3890
Gene Name
keratin 84
Gene Aliases
HB4, KRTHB4
UniGene
Hs.272336
Gene Chromosome Location
Chr.12: 52377665-52386977 on Build GRCh38
Transcripts
NM_033045.3 XM_011538335.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7428181 c.1068C>T p.Y356Y Chr.12: 52381370-52381370 on Build GRCh38
4512831 c.918C>T p.I306I Chr.12: 52381520-52381520 on Build GRCh38
1476666 c.991G>C p.D331H Chr.12: 52381447-52381447 on Build GRCh38
3376290 c.913-1G>T p.? Chr.12: 52381526-52381526 on Build GRCh38
328815 c.965T>A p.M322K Chr.12: 52381473-52381473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113837792 Chr.12: 52381392-52381392 on Build GRCh38
rs878860575 Chr.12: 52381432-52381432 on Build GRCh38
rs1010828735 Chr.12: 52381465-52381465 on Build GRCh38
rs139009727 Chr.12: 52381383-52381383 on Build GRCh38
rs748358930 Chr.12: 52381384-52381384 on Build GRCh38
rs758564487 Chr.12: 52381456-52381456 on Build GRCh38
rs867590669 Chr.12: 52381423-52381423 on Build GRCh38
rs754269484 Chr.12: 52381333-52381333 on Build GRCh38
rs766981703 Chr.12: 52381320-52381320 on Build GRCh38
rs374447110 Chr.12: 52381525-52381525 on Build GRCh38
rs144405225 Chr.12: 52381487-52381487 on Build GRCh38
rs555015096 Chr.12: 52381490-52381490 on Build GRCh38
rs781038880 Chr.12: 52381363-52381363 on Build GRCh38
rs370280302 Chr.12: 52381491-52381491 on Build GRCh38
rs779779148 Chr.12: 52381445-52381445 on Build GRCh38
rs756713899 Chr.12: 52381421-52381421 on Build GRCh38
rs758051534 Chr.12: 52381344-52381344 on Build GRCh38
rs763699382 Chr.12: 52381364-52381364 on Build GRCh38
rs114877532 Chr.12: 52381391-52381391 on Build GRCh38
rs749936071 Chr.12: 52381505-52381505 on Build GRCh38
rs748245138 Chr.12: 52381460-52381460 on Build GRCh38
rs769425885 Chr.12: 52381378-52381378 on Build GRCh38
rs781735766 Chr.12: 52381430-52381430 on Build GRCh38
rs999221955 Chr.12: 52381488-52381488 on Build GRCh38
rs373467704 Chr.12: 52381393-52381393 on Build GRCh38
rs775726115 Chr.12: 52381471-52381471 on Build GRCh38
rs751052503 Chr.12: 52381355-52381355 on Build GRCh38
rs765572778 Chr.12: 52381406-52381406 on Build GRCh38
rs966599327 Chr.12: 52381312-52381312 on Build GRCh38
rs776817362 Chr.12: 52381468-52381468 on Build GRCh38
rs759971351 Chr.12: 52381399-52381399 on Build GRCh38
rs777211528 Chr.12: 52381347-52381347 on Build GRCh38
rs1006699227 Chr.12: 52381353-52381353 on Build GRCh38
rs745691802 Chr.12: 52381377-52381377 on Build GRCh38
rs771092679 Chr.12: 52381467-52381467 on Build GRCh38
rs772334938 Chr.12: 52381387-52381387 on Build GRCh38
rs965413016 Chr.12: 52381375-52381375 on Build GRCh38
rs751070892 Chr.12: 52381415-52381415 on Build GRCh38
rs781366492 Chr.12: 52381393-52381393 on Build GRCh38
rs991579106 Chr.12: 52381464-52381464 on Build GRCh38
rs755277724 Chr.12: 52381356-52381356 on Build GRCh38
rs747106404 Chr.12: 52381463-52381463 on Build GRCh38
rs146876431 Chr.12: 52381398-52381398 on Build GRCh38
rs770334102 Chr.12: 52381470-52381470 on Build GRCh38
rs781128772 Chr.12: 52381440-52381440 on Build GRCh38
rs763322644 Chr.12: 52381479-52381479 on Build GRCh38
rs537689654 Chr.12: 52381459-52381459 on Build GRCh38
rs764975216 Chr.12: 52381335-52381335 on Build GRCh38
rs766271355 Chr.12: 52381520-52381520 on Build GRCh38
rs201789393 Chr.12: 52381343-52381343 on Build GRCh38
rs767063415 Chr.12: 52381503-52381503 on Build GRCh38
rs756047801 Chr.12: 52381441-52381441 on Build GRCh38
rs749096842 Chr.12: 52381453-52381453 on Build GRCh38
rs139738722 Chr.12: 52381462-52381462 on Build GRCh38
rs145321688 Chr.12: 52381511-52381511 on Build GRCh38
rs757195974 Chr.12: 52381358-52381358 on Build GRCh38
rs867746805 Chr.12: 52381461-52381461 on Build GRCh38
rs753078744 Chr.12: 52381408-52381408 on Build GRCh38
rs201514187 Chr.12: 52381334-52381334 on Build GRCh38
rs376898463 Chr.12: 52381414-52381414 on Build GRCh38
rs746407111 Chr.12: 52381469-52381469 on Build GRCh38
rs747198526 Chr.12: 52381311-52381311 on Build GRCh38

View Full Product Details