Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54847835-54848074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIR2DS4
Entrez Gene ID
3809
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
Gene Aliases
CD158I, KIR-2DS4, KIR1D, KIR2DS1, KIR412, KKA3, NKAT-8, NKAT8
UniGene
Hs.654608
Gene Chromosome Location
Chr.19: 54832676-54848569 on Build GRCh38
Transcripts
NM_001281971.1 NM_001281972.1 NM_012314.5

SNP Detail

SNP ID Chromosome Location
rs202174486 Chr.19: 54847931-54847931 on Build GRCh38
rs113552989 Chr.19: 54848049-54848049 on Build GRCh38
rs376072641 Chr.19: 54848030-54848030 on Build GRCh38
rs17173061 Chr.19: 54847989-54847989 on Build GRCh38
rs74623998 Chr.19: 54847971-54847971 on Build GRCh38
rs373068946 Chr.19: 54848020-54848020 on Build GRCh38
rs62122202 Chr.19: 54847949-54847949 on Build GRCh38
rs769385609 Chr.19: 54848014-54848014 on Build GRCh38
rs772562693 Chr.19: 54848028-54848028 on Build GRCh38
rs747537662 Chr.19: 54847992-54847992 on Build GRCh38
rs112305589 Chr.19: 54847916-54847916 on Build GRCh38
rs780649287 Chr.19: 54847986-54847986 on Build GRCh38
rs113532118 Chr.19: 54848017-54848017 on Build GRCh38
rs369652041 Chr.19: 54848006-54848006 on Build GRCh38
rs879086128 Chr.19: 54848046-54848046 on Build GRCh38
rs139032132 Chr.19: 54847981-54847981 on Build GRCh38

View Full Product Details