Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 91448664-91448921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPC5
Entrez Gene ID
2262
Gene Name
glypican 5
Gene Aliases
-
UniGene
Hs.655675
Gene Chromosome Location
Chr.13: 91398619-92867237 on Build GRCh38
Transcripts
NM_004466.5 XM_011521054.2 XM_011521055.2 XM_011521056.2 XM_011521057.2 XM_011521058.2 XM_011521059.2 XM_011521060.2 XM_017020435.1 XM_017020436.1 XM_017020437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208536 c.241G>A p.A81T Chr.13: 91448838-91448838 on Build GRCh38
1706891 c.169G>A p.D57N Chr.13: 91448766-91448766 on Build GRCh38
7369846 c.232C>A p.Q78K Chr.13: 91448829-91448829 on Build GRCh38
6074931 c.249G>T p.Q83H Chr.13: 91448846-91448846 on Build GRCh38
6881031 c.192A>T p.K64N Chr.13: 91448789-91448789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767268470 Chr.13: 91448895-91448895 on Build GRCh38
rs754305211 Chr.13: 91448816-91448816 on Build GRCh38
rs991147810 Chr.13: 91448698-91448698 on Build GRCh38
rs775250275 Chr.13: 91448796-91448796 on Build GRCh38
rs757294831 Chr.13: 91448841-91448841 on Build GRCh38
rs776573728 Chr.13: 91448775-91448775 on Build GRCh38
rs779425624 Chr.13: 91448746-91448746 on Build GRCh38
rs765904788 Chr.13: 91448830-91448830 on Build GRCh38
rs762782899 Chr.13: 91448805-91448805 on Build GRCh38
rs374581120 Chr.13: 91448747-91448747 on Build GRCh38
rs1023150476 Chr.13: 91448713-91448713 on Build GRCh38
rs778001489 Chr.13: 91448837-91448837 on Build GRCh38
rs769785386 Chr.13: 91448788-91448788 on Build GRCh38
rs573449648 Chr.13: 91448690-91448690 on Build GRCh38
rs925447067 Chr.13: 91448781-91448781 on Build GRCh38
rs935466194 Chr.13: 91448814-91448814 on Build GRCh38
rs759392232 Chr.13: 91448786-91448786 on Build GRCh38
rs770938385 Chr.13: 91448774-91448774 on Build GRCh38
rs755337270 Chr.13: 91448738-91448738 on Build GRCh38
rs968045189 Chr.13: 91448711-91448711 on Build GRCh38
rs147685128 Chr.13: 91448785-91448785 on Build GRCh38
rs772412094 Chr.13: 91448752-91448752 on Build GRCh38
rs775443187 Chr.13: 91448845-91448845 on Build GRCh38
rs774001219 Chr.13: 91448870-91448870 on Build GRCh38
rs371917437 Chr.13: 91448809-91448809 on Build GRCh38
rs777896189 Chr.13: 91448764-91448764 on Build GRCh38
rs368450803 Chr.13: 91448763-91448763 on Build GRCh38
rs375508754 Chr.13: 91448901-91448901 on Build GRCh38
rs568024128 Chr.13: 91448700-91448700 on Build GRCh38
rs190244359 Chr.13: 91448767-91448767 on Build GRCh38
rs747616494 Chr.13: 91448721-91448721 on Build GRCh38
rs528474256 Chr.13: 91448847-91448847 on Build GRCh38
rs559277707 Chr.13: 91448836-91448836 on Build GRCh38
rs761376470 Chr.13: 91448887-91448887 on Build GRCh38
rs747208506 Chr.13: 91448838-91448838 on Build GRCh38
rs778500286 Chr.13: 91448708-91448708 on Build GRCh38
rs527318537 Chr.13: 91448703-91448703 on Build GRCh38
rs745349116 Chr.13: 91448775-91448776 on Build GRCh38
rs933328176 Chr.13: 91448872-91448872 on Build GRCh38
rs142561779 Chr.13: 91448851-91448851 on Build GRCh38
rs754327954 Chr.13: 91448714-91448714 on Build GRCh38
rs761342792 Chr.13: 91448815-91448815 on Build GRCh38
rs773909450 Chr.13: 91448810-91448810 on Build GRCh38
rs201226040 Chr.13: 91448842-91448842 on Build GRCh38
rs549282768 Chr.13: 91448832-91448832 on Build GRCh38
rs543280491 Chr.13: 91448686-91448686 on Build GRCh38
rs529144873 Chr.13: 91448826-91448826 on Build GRCh38
rs142381235 Chr.13: 91448869-91448869 on Build GRCh38

View Full Product Details