Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23469937-23470210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGA2
Entrez Gene ID
23062
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
VEAR
UniGene
Hs.460336
Gene Chromosome Location
Chr.16: 23463542-23510494 on Build GRCh38
Transcripts
NM_015044.4 XM_017023074.1 XM_017023075.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6195143 c.1611T>C p.A537A Chr.16: 23470005-23470005 on Build GRCh38
6220818 c.1563G>A p.M521I Chr.16: 23470053-23470053 on Build GRCh38
7155988 c.1526C>T p.P509L Chr.16: 23470090-23470090 on Build GRCh38
79901 c.1596C>A p.I532I Chr.16: 23470020-23470020 on Build GRCh38
1563184 c.1487G>T p.G496V Chr.16: 23470129-23470129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753062374 Chr.16: 23470180-23470180 on Build GRCh38
rs751978774 Chr.16: 23470029-23470029 on Build GRCh38
rs760553146 Chr.16: 23470129-23470129 on Build GRCh38
rs745823263 Chr.16: 23470130-23470130 on Build GRCh38
rs767623820 Chr.16: 23469993-23469993 on Build GRCh38
rs775368972 Chr.16: 23470113-23470113 on Build GRCh38
rs767889220 Chr.16: 23470173-23470173 on Build GRCh38
rs756592274 Chr.16: 23470045-23470045 on Build GRCh38
rs568617546 Chr.16: 23470022-23470022 on Build GRCh38
rs748797331 Chr.16: 23470065-23470065 on Build GRCh38
rs774323871 Chr.16: 23469971-23469971 on Build GRCh38
rs182464792 Chr.16: 23469986-23469986 on Build GRCh38
rs760596835 Chr.16: 23470009-23470009 on Build GRCh38
rs766654195 Chr.16: 23470160-23470160 on Build GRCh38
rs139811732 Chr.16: 23470053-23470053 on Build GRCh38
rs772000231 Chr.16: 23470102-23470102 on Build GRCh38
rs566563362 Chr.16: 23470016-23470016 on Build GRCh38
rs778698998 Chr.16: 23470098-23470098 on Build GRCh38
rs922640002 Chr.16: 23470101-23470101 on Build GRCh38
rs201439823 Chr.16: 23470099-23470099 on Build GRCh38
rs149792096 Chr.16: 23470156-23470156 on Build GRCh38
rs769436325 Chr.16: 23470145-23470145 on Build GRCh38
rs551493440 Chr.16: 23470071-23470071 on Build GRCh38
rs775321784 Chr.16: 23470176-23470176 on Build GRCh38
rs967971570 Chr.16: 23470040-23470040 on Build GRCh38
rs1034705929 Chr.16: 23469964-23469964 on Build GRCh38
rs777175077 Chr.16: 23470055-23470055 on Build GRCh38
rs111796012 Chr.16: 23470061-23470061 on Build GRCh38
rs759694967 Chr.16: 23470168-23470168 on Build GRCh38
rs775567103 Chr.16: 23469996-23469996 on Build GRCh38
rs202188100 Chr.16: 23470135-23470135 on Build GRCh38
rs371782021 Chr.16: 23470161-23470161 on Build GRCh38
rs765246769 Chr.16: 23470019-23470019 on Build GRCh38
rs770573317 Chr.16: 23469965-23469965 on Build GRCh38
rs774314067 Chr.16: 23470151-23470151 on Build GRCh38
rs200357653 Chr.16: 23470020-23470020 on Build GRCh38
rs992901147 Chr.16: 23470025-23470025 on Build GRCh38
rs367720663 Chr.16: 23470044-23470044 on Build GRCh38
rs865878713 Chr.16: 23470182-23470182 on Build GRCh38
rs755330227 Chr.16: 23470039-23470039 on Build GRCh38

View Full Product Details