Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93110165-93110414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTF2
Entrez Gene ID
22823
Gene Name
metal response element binding transcription factor 2
Gene Aliases
M96, PCL2, TDRD19A, dJ976O13.2
UniGene
Hs.31016
Gene Chromosome Location
Chr.1: 93079219-93139081 on Build GRCh38
Transcripts
NM_001164391.1 NM_001164392.1 NM_001164393.1 NM_007358.3 XM_011541016.1 XM_011541017.2 XM_011541019.1 XM_017000676.1 XM_017000677.1 XM_017000678.1 XM_017000679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
465101 c.182T>C p.F61S Chr.1: 93110406-93110406 on Build GRCh38
1602805 c.68A>T p.K23M Chr.1: 93110292-93110292 on Build GRCh38
4703528 c.15_16insG p.A7fs*3 Chr.1: 93110239-93110240 on Build GRCh38
282909 c.69G>T p.K23N Chr.1: 93110293-93110293 on Build GRCh38
4703529 c.76A>C p.T26P Chr.1: 93110300-93110300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750955831 Chr.1: 93110265-93110265 on Build GRCh38
rs769799253 Chr.1: 93110224-93110224 on Build GRCh38
rs760932337 Chr.1: 93110364-93110364 on Build GRCh38
rs137905512 Chr.1: 93110273-93110273 on Build GRCh38
rs140305129 Chr.1: 93110257-93110257 on Build GRCh38
rs143820772 Chr.1: 93110276-93110276 on Build GRCh38
rs777671958 Chr.1: 93110290-93110290 on Build GRCh38
rs754440392 Chr.1: 93110279-93110279 on Build GRCh38
rs767694863 Chr.1: 93110343-93110343 on Build GRCh38
rs761980174 Chr.1: 93110332-93110332 on Build GRCh38
rs770953603 Chr.1: 93110309-93110309 on Build GRCh38
rs781395811 Chr.1: 93110355-93110355 on Build GRCh38
rs778633481 Chr.1: 93110280-93110280 on Build GRCh38
rs758291787 Chr.1: 93110284-93110284 on Build GRCh38
rs941890613 Chr.1: 93110313-93110313 on Build GRCh38
rs189234488 Chr.1: 93110283-93110283 on Build GRCh38
rs774487835 Chr.1: 93110255-93110255 on Build GRCh38
rs773296018 Chr.1: 93110199-93110199 on Build GRCh38
rs762923381 Chr.1: 93110241-93110241 on Build GRCh38
rs183737932 Chr.1: 93110261-93110261 on Build GRCh38
rs768559181 Chr.1: 93110325-93110325 on Build GRCh38
rs749454906 Chr.1: 93110317-93110317 on Build GRCh38
rs574583820 Chr.1: 93110373-93110373 on Build GRCh38
rs1054951816 Chr.1: 93110334-93110334 on Build GRCh38
rs776422711 Chr.1: 93110311-93110311 on Build GRCh38
rs760640594 Chr.1: 93110205-93110205 on Build GRCh38
rs889679212 Chr.1: 93110368-93110368 on Build GRCh38
rs756684732 Chr.1: 93110266-93110266 on Build GRCh38
rs766773407 Chr.1: 93110267-93110267 on Build GRCh38
rs774575875 Chr.1: 93110326-93110326 on Build GRCh38
rs764436816 Chr.1: 93110247-93110247 on Build GRCh38
rs746773581 Chr.1: 93110295-93110295 on Build GRCh38
rs775682782 Chr.1: 93110236-93110236 on Build GRCh38
rs983569243 Chr.1: 93110304-93110304 on Build GRCh38
rs773616090 Chr.1: 93110353-93110353 on Build GRCh38

View Full Product Details