Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54509537-54509808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCKAP1L
Entrez Gene ID
3071
Gene Name
NCK associated protein 1 like
Gene Aliases
HEM1
UniGene
Hs.182014
Gene Chromosome Location
Chr.12: 54497711-54543115 on Build GRCh38
Transcripts
NM_001184976.1 NM_005337.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
162828 c.547T>A p.Y183N Chr.12: 54509709-54509709 on Build GRCh38
6722964 c.573A>G p.T191T Chr.12: 54509735-54509735 on Build GRCh38
3871972 c.589C>T p.H197Y Chr.12: 54509751-54509751 on Build GRCh38
941155 c.525T>C p.R175R Chr.12: 54509687-54509687 on Build GRCh38
3462823 c.577G>A p.E193K Chr.12: 54509739-54509739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754013750 Chr.12: 54509723-54509723 on Build GRCh38
rs113374416 Chr.12: 54509705-54509705 on Build GRCh38
rs775195882 Chr.12: 54509677-54509677 on Build GRCh38
rs200735062 Chr.12: 54509660-54509660 on Build GRCh38
rs778722766 Chr.12: 54509745-54509745 on Build GRCh38
rs562610374 Chr.12: 54509691-54509691 on Build GRCh38
rs991985224 Chr.12: 54509714-54509714 on Build GRCh38
rs867657709 Chr.12: 54509658-54509658 on Build GRCh38
rs917064361 Chr.12: 54509716-54509716 on Build GRCh38
rs201962488 Chr.12: 54509623-54509623 on Build GRCh38
rs766973721 Chr.12: 54509715-54509715 on Build GRCh38
rs373702631 Chr.12: 54509605-54509605 on Build GRCh38
rs756714425 Chr.12: 54509707-54509707 on Build GRCh38
rs180951280 Chr.12: 54509686-54509686 on Build GRCh38
rs898694580 Chr.12: 54509601-54509601 on Build GRCh38
rs901357142 Chr.12: 54509566-54509566 on Build GRCh38
rs201133011 Chr.12: 54509755-54509755 on Build GRCh38
rs752974183 Chr.12: 54509753-54509753 on Build GRCh38
rs1039881438 Chr.12: 54509593-54509593 on Build GRCh38
rs779059689 Chr.12: 54509741-54509741 on Build GRCh38
rs759022603 Chr.12: 54509665-54509665 on Build GRCh38
rs764938318 Chr.12: 54509669-54509669 on Build GRCh38
rs951343275 Chr.12: 54509605-54509605 on Build GRCh38
rs958744621 Chr.12: 54509713-54509713 on Build GRCh38
rs369137316 Chr.12: 54509732-54509732 on Build GRCh38
rs1026177679 Chr.12: 54509646-54509646 on Build GRCh38
rs369935244 Chr.12: 54509636-54509636 on Build GRCh38
rs777655016 Chr.12: 54509776-54509776 on Build GRCh38
rs540179913 Chr.12: 54509621-54509621 on Build GRCh38
rs998420242 Chr.12: 54509574-54509574 on Build GRCh38
rs746950075 Chr.12: 54509626-54509626 on Build GRCh38
rs942833362 Chr.12: 54509579-54509579 on Build GRCh38

View Full Product Details