Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113169326-113169581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP15
Entrez Gene ID
23307
Gene Name
FK506 binding protein 15
Gene Aliases
FKBP133, KIAA0674, PPP1R76
UniGene
Hs.522351
Gene Chromosome Location
Chr.9: 113165036-113221361 on Build GRCh38
Transcripts
NM_015258.1 XM_006717018.2 XM_006717019.1 XM_017014552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5086268 c.3408G>A p.G1136G Chr.9: 113169376-113169376 on Build GRCh38
3982703 c.3286C>T p.L1096L Chr.9: 113169423-113169423 on Build GRCh38
7080704 c.3430C>G p.Q1144E Chr.9: 113169354-113169354 on Build GRCh38
3653168 c.3234C>T p.V1078V Chr.9: 113169475-113169475 on Build GRCh38
244537 c.3196C>A p.P1066T Chr.9: 113169513-113169513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769859688 Chr.9: 113169373-113169373 on Build GRCh38
rs759345947 Chr.9: 113169398-113169398 on Build GRCh38
rs750024466 Chr.9: 113169534-113169534 on Build GRCh38
rs774647096 Chr.9: 113169477-113169477 on Build GRCh38
rs771860786 Chr.9: 113169495-113169495 on Build GRCh38
rs369380780 Chr.9: 113169409-113169409 on Build GRCh38
rs3810909 Chr.9: 113169412-113169412 on Build GRCh38
rs373180661 Chr.9: 113169388-113169388 on Build GRCh38
rs760566242 Chr.9: 113169501-113169501 on Build GRCh38
rs771270962 Chr.9: 113169395-113169395 on Build GRCh38
rs756543848 Chr.9: 113169446-113169446 on Build GRCh38
rs916831500 Chr.9: 113169393-113169393 on Build GRCh38
rs748591067 Chr.9: 113169442-113169442 on Build GRCh38
rs748612293 Chr.9: 113169372-113169372 on Build GRCh38
rs61748833 Chr.9: 113169504-113169504 on Build GRCh38
rs781427576 Chr.9: 113169424-113169424 on Build GRCh38
rs764737398 Chr.9: 113169513-113169513 on Build GRCh38
rs764142393 Chr.9: 113169406-113169406 on Build GRCh38
rs1037445074 Chr.9: 113169377-113169377 on Build GRCh38
rs755397372 Chr.9: 113169360-113169360 on Build GRCh38
rs572313532 Chr.9: 113169402-113169402 on Build GRCh38
rs746114964 Chr.9: 113169481-113169481 on Build GRCh38
rs3810910 Chr.9: 113169423-113169423 on Build GRCh38
rs775116359 Chr.9: 113169496-113169496 on Build GRCh38
rs781684581 Chr.9: 113169364-113169364 on Build GRCh38
rs764858932 Chr.9: 113169413-113169413 on Build GRCh38
rs551811341 Chr.9: 113169415-113169415 on Build GRCh38
rs148550803 Chr.9: 113169466-113169466 on Build GRCh38
rs749364046 Chr.9: 113169457-113169457 on Build GRCh38
rs760552921 Chr.9: 113169403-113169403 on Build GRCh38
rs773140343 Chr.9: 113169389-113169389 on Build GRCh38
rs778488499 Chr.9: 113169450-113169450 on Build GRCh38
rs757961097 Chr.9: 113169536-113169536 on Build GRCh38
rs767220150 Chr.9: 113169399-113169399 on Build GRCh38
rs889720622 Chr.9: 113169487-113169487 on Build GRCh38
rs995174390 Chr.9: 113169455-113169455 on Build GRCh38
rs185760376 Chr.9: 113169394-113169394 on Build GRCh38
rs780454081 Chr.9: 113169552-113169552 on Build GRCh38
rs766107320 Chr.9: 113169545-113169545 on Build GRCh38
rs543834605 Chr.9: 113169510-113169510 on Build GRCh38

View Full Product Details