Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98354124-98354361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5K4
Entrez Gene ID
403278
Gene Name
olfactory receptor family 5 subfamily K member 4
Gene Aliases
-
UniGene
Hs.554710
Gene Chromosome Location
Chr.3: 98353854-98354819 on Build GRCh38
Transcripts
NM_001005517.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1049188 c.294G>A p.M98I Chr.3: 98354147-98354147 on Build GRCh38
370100 c.365G>T p.R122L Chr.3: 98354218-98354218 on Build GRCh38
1049189 c.427C>T p.R143W Chr.3: 98354280-98354280 on Build GRCh38
4996378 c.303T>A p.F101L Chr.3: 98354156-98354156 on Build GRCh38
2950363 c.291T>C p.C97C Chr.3: 98354144-98354144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753910291 Chr.3: 98354264-98354264 on Build GRCh38
rs764544901 Chr.3: 98354180-98354180 on Build GRCh38
rs202129325 Chr.3: 98354295-98354295 on Build GRCh38
rs764445513 Chr.3: 98354244-98354244 on Build GRCh38
rs750716290 Chr.3: 98354258-98354258 on Build GRCh38
rs527631755 Chr.3: 98354246-98354246 on Build GRCh38
rs373649244 Chr.3: 98354280-98354280 on Build GRCh38
rs570495811 Chr.3: 98354270-98354270 on Build GRCh38
rs756366710 Chr.3: 98354261-98354261 on Build GRCh38
rs757509598 Chr.3: 98354196-98354196 on Build GRCh38
rs972146706 Chr.3: 98354274-98354274 on Build GRCh38
rs762142876 Chr.3: 98354249-98354249 on Build GRCh38
rs139091477 Chr.3: 98354330-98354330 on Build GRCh38
rs145946605 Chr.3: 98354292-98354292 on Build GRCh38
rs140109788 Chr.3: 98354221-98354221 on Build GRCh38
rs189244881 Chr.3: 98354204-98354204 on Build GRCh38
rs143150948 Chr.3: 98354293-98354293 on Build GRCh38
rs745548353 Chr.3: 98354284-98354284 on Build GRCh38
rs770676811 Chr.3: 98354277-98354277 on Build GRCh38
rs371406644 Chr.3: 98354263-98354263 on Build GRCh38
rs763466783 Chr.3: 98354238-98354238 on Build GRCh38
rs547691527 Chr.3: 98354269-98354269 on Build GRCh38
rs753250614 Chr.3: 98354165-98354165 on Build GRCh38
rs756311665 Chr.3: 98354201-98354201 on Build GRCh38
rs748867723 Chr.3: 98354294-98354294 on Build GRCh38
rs112311698 Chr.3: 98354262-98354262 on Build GRCh38
rs767902546 Chr.3: 98354256-98354256 on Build GRCh38
rs562966935 Chr.3: 98354174-98354174 on Build GRCh38
rs774770984 Chr.3: 98354315-98354315 on Build GRCh38
rs368785402 Chr.3: 98354156-98354156 on Build GRCh38
rs769331138 Chr.3: 98354227-98354227 on Build GRCh38
rs1028509379 Chr.3: 98354184-98354184 on Build GRCh38
rs1024135691 Chr.3: 98354218-98354218 on Build GRCh38
rs531428374 Chr.3: 98354202-98354202 on Build GRCh38
rs774937744 Chr.3: 98354233-98354233 on Build GRCh38
rs185693146 Chr.3: 98354200-98354200 on Build GRCh38
rs144521594 Chr.3: 98354217-98354217 on Build GRCh38
rs146616455 Chr.3: 98354209-98354209 on Build GRCh38
rs762350440 Chr.3: 98354319-98354319 on Build GRCh38
rs759161767 Chr.3: 98354223-98354223 on Build GRCh38
rs751814650 Chr.3: 98354183-98354183 on Build GRCh38

View Full Product Details