Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42160231-42160466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH4
Entrez Gene ID
23415
Gene Name
potassium voltage-gated channel subfamily H member 4
Gene Aliases
BEC2, ELK1, Kv12.3
UniGene
Hs.304081
Gene Chromosome Location
Chr.17: 42156892-42181278 on Build GRCh38
Transcripts
NM_012285.2 XM_017024400.1 XM_017024401.1 XM_017024402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6016640 c.2741C>T p.P914L Chr.17: 42160353-42160353 on Build GRCh38
6523537 c.2763C>T p.S921S Chr.17: 42160331-42160331 on Build GRCh38
5386381 c.2698G>A p.E900K Chr.17: 42160396-42160396 on Build GRCh38
6464860 c.2860G>T p.A954S Chr.17: 42160234-42160234 on Build GRCh38
5747852 c.2659A>G p.I887V Chr.17: 42160435-42160435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147972805 Chr.17: 42160373-42160373 on Build GRCh38
rs138917401 Chr.17: 42160331-42160331 on Build GRCh38
rs769242830 Chr.17: 42160370-42160370 on Build GRCh38
rs267604884 Chr.17: 42160252-42160252 on Build GRCh38
rs201119846 Chr.17: 42160306-42160306 on Build GRCh38
rs760437588 Chr.17: 42160390-42160390 on Build GRCh38
rs531842015 Chr.17: 42160362-42160362 on Build GRCh38
rs1016814011 Chr.17: 42160270-42160270 on Build GRCh38
rs761111147 Chr.17: 42160413-42160413 on Build GRCh38
rs373149737 Chr.17: 42160349-42160349 on Build GRCh38
rs759077847 Chr.17: 42160406-42160406 on Build GRCh38
rs774787808 Chr.17: 42160288-42160288 on Build GRCh38
rs267604885 Chr.17: 42160279-42160279 on Build GRCh38
rs200945829 Chr.17: 42160354-42160354 on Build GRCh38
rs762294742 Chr.17: 42160290-42160290 on Build GRCh38
rs143838785 Chr.17: 42160372-42160372 on Build GRCh38
rs765112953 Chr.17: 42160415-42160415 on Build GRCh38
rs750142478 Chr.17: 42160312-42160312 on Build GRCh38
rs969676719 Chr.17: 42160433-42160433 on Build GRCh38
rs769747845 Chr.17: 42160346-42160346 on Build GRCh38
rs778828449 Chr.17: 42160335-42160335 on Build GRCh38
rs199559049 Chr.17: 42160272-42160272 on Build GRCh38
rs934652542 Chr.17: 42160421-42160421 on Build GRCh38
rs767226950 Chr.17: 42160303-42160303 on Build GRCh38
rs145104718 Chr.17: 42160330-42160330 on Build GRCh38
rs753279380 Chr.17: 42160399-42160399 on Build GRCh38
rs377595778 Chr.17: 42160380-42160380 on Build GRCh38
rs760076806 Chr.17: 42160322-42160322 on Build GRCh38
rs1024598233 Chr.17: 42160428-42160428 on Build GRCh38
rs561074750 Chr.17: 42160398-42160398 on Build GRCh38
rs752682354 Chr.17: 42160417-42160417 on Build GRCh38
rs149002026 Chr.17: 42160342-42160342 on Build GRCh38
rs765983804 Chr.17: 42160329-42160329 on Build GRCh38
rs375831329 Chr.17: 42160337-42160337 on Build GRCh38
rs758213425 Chr.17: 42160340-42160340 on Build GRCh38
rs772738223 Chr.17: 42160389-42160389 on Build GRCh38

View Full Product Details