Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10396113-10396386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH8
Entrez Gene ID
4626
Gene Name
myosin heavy chain 8
Gene Aliases
DA7, MyHC-peri, MyHC-pn, gtMHC-F
UniGene
Hs.700484
Gene Chromosome Location
Chr.17: 10390325-10421950 on Build GRCh38
Transcripts
NM_002472.2 NR_125367.1

Gene Symbol
MYHAS
Entrez Gene ID
100128560
Gene Name
myosin heavy chain gene cluster antisense RNA
Gene Aliases
-
UniGene
Hs.661174
Gene Chromosome Location
Chr.17: 10383132-10625540 on Build GRCh38
Transcripts
NM_002472.2 NR_125367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4605422 c.4606G>A p.V1536I Chr.17: 10396377-10396377 on Build GRCh38
3712199 c.4602G>C p.K1534N Chr.17: 10396381-10396381 on Build GRCh38
5385632 c.4624G>A p.E1542K Chr.17: 10396359-10396359 on Build GRCh38
3969793 c.4603C>T p.Q1535* Chr.17: 10396380-10396380 on Build GRCh38
3513632 c.4617G>A p.E1539E Chr.17: 10396366-10396366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903136232 Chr.17: 10396146-10396146 on Build GRCh38
rs943142697 Chr.17: 10396220-10396220 on Build GRCh38
rs567077328 Chr.17: 10396147-10396147 on Build GRCh38
rs998784835 Chr.17: 10396161-10396166 on Build GRCh38
rs755552897 Chr.17: 10396311-10396311 on Build GRCh38
rs954373649 Chr.17: 10396235-10396235 on Build GRCh38
rs1017389848 Chr.17: 10396309-10396309 on Build GRCh38
rs35813479 Chr.17: 10396179-10396180 on Build GRCh38
rs770256868 Chr.17: 10396318-10396318 on Build GRCh38
rs989856088 Chr.17: 10396140-10396140 on Build GRCh38
rs751587199 Chr.17: 10396294-10396294 on Build GRCh38
rs779989441 Chr.17: 10396285-10396285 on Build GRCh38
rs199814495 Chr.17: 10396351-10396351 on Build GRCh38
rs914309209 Chr.17: 10396209-10396209 on Build GRCh38
rs560559948 Chr.17: 10396176-10396189 on Build GRCh38
rs1029967997 Chr.17: 10396205-10396205 on Build GRCh38
rs777530660 Chr.17: 10396322-10396322 on Build GRCh38
rs146599852 Chr.17: 10396246-10396246 on Build GRCh38
rs781746939 Chr.17: 10396313-10396313 on Build GRCh38
rs748880308 Chr.17: 10396350-10396350 on Build GRCh38
rs200656694 Chr.17: 10396315-10396315 on Build GRCh38

View Full Product Details