Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 50376602-50376827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF1
Entrez Gene ID
10320
Gene Name
IKAROS family zinc finger 1
Gene Aliases
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1
UniGene
Hs.435949
Gene Chromosome Location
Chr.7: 50303453-50405101 on Build GRCh38
Transcripts
NM_001220765.2 NM_001220767.2 NM_001220768.2 NM_001220770.2 NM_001220771.2 NM_001291837.1 NM_001291838.1 NM_001291839.1 NM_001291840.1 NM_001291841.1 NM_001291842.1 NM_001291843.1 NM_001291844.1 NM_006060.5 XM_011515058.2 XM_011515059.2 XM_011515060.2 XM_011515061.2 XM_011515062.2 XM_011515063.2 XM_011515064.2 XM_011515065.2 XM_011515066.2 XM_011515067.2 XM_011515068.2 XM_011515069.2 XM_011515070.2 XM_011515071.2 XM_011515072.2 XM_011515073.2 XM_011515074.2 XM_011515075.2 XM_011515076.2 XM_011515077.2 XM_011515078.2 XM_017011667.1 XM_017011668.1 XM_017011669.1 XM_017011670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
87302 c.308_309ins23 p.R111fs*15 Chr.7: 50376680-50376681 on Build GRCh38
85911 c.161-?_1560+?del p.? Chr.7: 50376533-50400627 on Build GRCh38
85912 c.1-?_850+?del p.? Chr.7: 50342934-50391863 on Build GRCh38
85915 c.161-?_850+?del p.? Chr.7: 50376533-50391863 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs868197423 Chr.7: 50376688-50376688 on Build GRCh38
rs766101392 Chr.7: 50376699-50376699 on Build GRCh38
rs549930725 Chr.7: 50376689-50376689 on Build GRCh38
rs1032782282 Chr.7: 50376631-50376631 on Build GRCh38
rs185632810 Chr.7: 50376714-50376714 on Build GRCh38
rs764780164 Chr.7: 50376687-50376687 on Build GRCh38
rs771632614 Chr.7: 50376628-50376628 on Build GRCh38
rs942938472 Chr.7: 50376652-50376652 on Build GRCh38
rs767627243 Chr.7: 50376756-50376756 on Build GRCh38
rs764847531 Chr.7: 50376770-50376770 on Build GRCh38
rs777855187 Chr.7: 50376804-50376804 on Build GRCh38
rs746327678 Chr.7: 50376636-50376636 on Build GRCh38
rs761574077 Chr.7: 50376686-50376686 on Build GRCh38
rs756418516 Chr.7: 50376800-50376800 on Build GRCh38
rs759433474 Chr.7: 50376721-50376721 on Build GRCh38
rs776248281 Chr.7: 50376684-50376684 on Build GRCh38
rs776572250 Chr.7: 50376677-50376677 on Build GRCh38
rs775004468 Chr.7: 50376635-50376635 on Build GRCh38

View Full Product Details