Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100888626-100888893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL2
Entrez Gene ID
2135
Gene Name
exostosin like glycosyltransferase 2
Gene Aliases
EXTR2
UniGene
Hs.357637
Gene Chromosome Location
Chr.1: 100872372-100895411 on Build GRCh38
Transcripts
NM_001033025.2 NM_001261440.1 NM_001261441.1 NM_001261442.1 NM_001439.3 NR_048570.1 XM_005270621.1 XM_011540994.2 XM_011540995.2 XM_011540996.2 XM_017000651.1

SNP Detail

SNP ID Chromosome Location
rs767340271 Chr.1: 100888652-100888652 on Build GRCh38
rs565523516 Chr.1: 100888767-100888767 on Build GRCh38
rs748051556 Chr.1: 100888724-100888724 on Build GRCh38
rs755735504 Chr.1: 100888673-100888673 on Build GRCh38
rs373028355 Chr.1: 100888770-100888770 on Build GRCh38
rs373536267 Chr.1: 100888847-100888847 on Build GRCh38
rs775904520 Chr.1: 100888727-100888727 on Build GRCh38
rs953827967 Chr.1: 100888792-100888792 on Build GRCh38
rs1046950633 Chr.1: 100888679-100888679 on Build GRCh38
rs768548911 Chr.1: 100888844-100888844 on Build GRCh38
rs376269400 Chr.1: 100888820-100888820 on Build GRCh38
rs977688773 Chr.1: 100888804-100888804 on Build GRCh38
rs749372270 Chr.1: 100888754-100888754 on Build GRCh38
rs77649301 Chr.1: 100888830-100888830 on Build GRCh38
rs767940159 Chr.1: 100888765-100888765 on Build GRCh38
rs766003550 Chr.1: 100888778-100888778 on Build GRCh38
rs976289240 Chr.1: 100888838-100888838 on Build GRCh38
rs768784866 Chr.1: 100888757-100888757 on Build GRCh38
rs754815967 Chr.1: 100888745-100888745 on Build GRCh38
rs374861571 Chr.1: 100888760-100888760 on Build GRCh38
rs886953612 Chr.1: 100888680-100888680 on Build GRCh38
rs74772581 Chr.1: 100888677-100888677 on Build GRCh38
rs755750928 Chr.1: 100888704-100888704 on Build GRCh38
rs778818110 Chr.1: 100888718-100888718 on Build GRCh38
rs1015543634 Chr.1: 100888729-100888729 on Build GRCh38
rs914187042 Chr.1: 100888839-100888839 on Build GRCh38
rs1019845745 Chr.1: 100888684-100888684 on Build GRCh38
rs1008089727 Chr.1: 100888682-100888682 on Build GRCh38
rs200957907 Chr.1: 100888768-100888768 on Build GRCh38
rs753325088 Chr.1: 100888707-100888707 on Build GRCh38
rs764587983 Chr.1: 100888777-100888778 on Build GRCh38
rs968336407 Chr.1: 100888803-100888803 on Build GRCh38
rs376846404 Chr.1: 100888706-100888706 on Build GRCh38
rs758284425 Chr.1: 100888732-100888732 on Build GRCh38
rs76722689 Chr.1: 100888808-100888808 on Build GRCh38
rs111669428 Chr.1: 100888855-100888855 on Build GRCh38
rs115405427 Chr.1: 100888737-100888737 on Build GRCh38
rs996592778 Chr.1: 100888748-100888748 on Build GRCh38
rs556632131 Chr.1: 100888822-100888822 on Build GRCh38
rs551290904 Chr.1: 100888654-100888654 on Build GRCh38
rs746813732 Chr.1: 100888742-100888742 on Build GRCh38
rs571799879 Chr.1: 100888715-100888715 on Build GRCh38
rs995384689 Chr.1: 100888665-100888665 on Build GRCh38
rs554399525 Chr.1: 100888758-100888758 on Build GRCh38

View Full Product Details