Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154022253-154022488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP210L
Entrez Gene ID
91181
Gene Name
nucleoporin 210 like
Gene Aliases
-
UniGene
Hs.67639
Gene Chromosome Location
Chr.1: 153992582-154155116 on Build GRCh38
Transcripts
NM_001159484.1 NM_207308.2 XM_011510122.1 XM_011510123.1 XM_011510124.1 XM_017002788.1 XM_017002789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
675916 c.4354G>C p.A1452P Chr.1: 154022288-154022288 on Build GRCh38
6432655 c.4388G>T p.W1463L Chr.1: 154022254-154022254 on Build GRCh38
4891445 c.4344C>A p.Y1448* Chr.1: 154022298-154022298 on Build GRCh38
3731703 c.4341T>C p.T1447T Chr.1: 154022301-154022301 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747800251 Chr.1: 154022384-154022384 on Build GRCh38
rs751411024 Chr.1: 154022374-154022374 on Build GRCh38
rs769111561 Chr.1: 154022411-154022411 on Build GRCh38
rs958145263 Chr.1: 154022274-154022274 on Build GRCh38
rs976636669 Chr.1: 154022356-154022356 on Build GRCh38
rs759987835 Chr.1: 154022296-154022296 on Build GRCh38
rs754789226 Chr.1: 154022376-154022376 on Build GRCh38
rs775878129 Chr.1: 154022314-154022314 on Build GRCh38
rs369671902 Chr.1: 154022272-154022272 on Build GRCh38
rs916624548 Chr.1: 154022395-154022395 on Build GRCh38
rs766379466 Chr.1: 154022362-154022362 on Build GRCh38
rs760924393 Chr.1: 154022330-154022330 on Build GRCh38
rs961335309 Chr.1: 154022329-154022329 on Build GRCh38
rs932496382 Chr.1: 154022440-154022440 on Build GRCh38
rs767957235 Chr.1: 154022313-154022313 on Build GRCh38
rs764274510 Chr.1: 154022340-154022340 on Build GRCh38
rs780904179 Chr.1: 154022381-154022381 on Build GRCh38
rs754016883 Chr.1: 154022349-154022349 on Build GRCh38
rs774701850 Chr.1: 154022295-154022295 on Build GRCh38
rs757260813 Chr.1: 154022353-154022353 on Build GRCh38
rs771529831 Chr.1: 154022284-154022284 on Build GRCh38

View Full Product Details