Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36942533-36942782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHX1
Entrez Gene ID
3975
Gene Name
LIM homeobox 1
Gene Aliases
LIM-1, LIM1
UniGene
Hs.443727
Gene Chromosome Location
Chr.17: 36937475-36944615 on Build GRCh38
Transcripts
NM_005568.4

SNP Detail

SNP ID Chromosome Location
rs893639065 Chr.17: 36942601-36942601 on Build GRCh38
rs912733730 Chr.17: 36942698-36942698 on Build GRCh38
rs987273307 Chr.17: 36942689-36942689 on Build GRCh38
rs933288966 Chr.17: 36942557-36942557 on Build GRCh38
rs556078877 Chr.17: 36942704-36942704 on Build GRCh38
rs749084444 Chr.17: 36942705-36942705 on Build GRCh38
rs1003579277 Chr.17: 36942635-36942635 on Build GRCh38
rs766878151 Chr.17: 36942741-36942741 on Build GRCh38
rs767076022 Chr.17: 36942731-36942731 on Build GRCh38
rs761558885 Chr.17: 36942719-36942719 on Build GRCh38
rs548348928 Chr.17: 36942610-36942610 on Build GRCh38
rs570337695 Chr.17: 36942699-36942699 on Build GRCh38
rs533711091 Chr.17: 36942672-36942672 on Build GRCh38
rs1051791250 Chr.17: 36942563-36942563 on Build GRCh38
rs906466719 Chr.17: 36942618-36942618 on Build GRCh38
rs922236717 Chr.17: 36942555-36942555 on Build GRCh38
rs989033854 Chr.17: 36942703-36942703 on Build GRCh38
rs963867352 Chr.17: 36942717-36942717 on Build GRCh38
rs772833109 Chr.17: 36942735-36942735 on Build GRCh38
rs951059938 Chr.17: 36942685-36942685 on Build GRCh38
rs539331458 Chr.17: 36942702-36942702 on Build GRCh38
rs754324037 Chr.17: 36942743-36942743 on Build GRCh38
rs774104700 Chr.17: 36942714-36942714 on Build GRCh38
rs28502075 Chr.17: 36942651-36942651 on Build GRCh38
rs1045374384 Chr.17: 36942615-36942615 on Build GRCh38
rs760168771 Chr.17: 36942740-36942740 on Build GRCh38
rs553570625 Chr.17: 36942683-36942683 on Build GRCh38
rs569268773 Chr.17: 36942564-36942564 on Build GRCh38
rs1035923712 Chr.17: 36942572-36942572 on Build GRCh38
rs923989426 Chr.17: 36942573-36942573 on Build GRCh38
rs768354752 Chr.17: 36942708-36942708 on Build GRCh38
rs1053834154 Chr.17: 36942580-36942580 on Build GRCh38
rs146331094 Chr.17: 36942654-36942654 on Build GRCh38
rs191105529 Chr.17: 36942576-36942576 on Build GRCh38
rs775026965 Chr.17: 36942703-36942703 on Build GRCh38

View Full Product Details