Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35250210-35250441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSR
Entrez Gene ID
51599
Gene Name
lipolysis stimulated lipoprotein receptor
Gene Aliases
ILDR3, LISCH7
UniGene
Hs.466507
Gene Chromosome Location
Chr.19: 35248656-35267964 on Build GRCh38
Transcripts
NM_001260489.1 NM_001260490.1 NM_015925.6 NM_205834.3 NM_205835.3 XM_005258980.1 XM_005258982.1 XM_011527026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5885852 c.254-8delG p.? Chr.19: 35250307-35250307 on Build GRCh38
1286020 c.271C>A p.Q91K Chr.19: 35250332-35250332 on Build GRCh38
4076951 c.333T>C p.C111C Chr.19: 35250394-35250394 on Build GRCh38
3532460 c.287A>G p.N96S Chr.19: 35250348-35250348 on Build GRCh38
3532459 c.278C>T p.T93I Chr.19: 35250339-35250339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037903700 Chr.19: 35250302-35250302 on Build GRCh38
rs148172260 Chr.19: 35250411-35250411 on Build GRCh38
rs777277378 Chr.19: 35250271-35250271 on Build GRCh38
rs899396862 Chr.19: 35250337-35250337 on Build GRCh38
rs202026383 Chr.19: 35250421-35250421 on Build GRCh38
rs755992438 Chr.19: 35250353-35250353 on Build GRCh38
rs760057788 Chr.19: 35250272-35250272 on Build GRCh38
rs993813274 Chr.19: 35250312-35250312 on Build GRCh38
rs770385789 Chr.19: 35250406-35250406 on Build GRCh38
rs751281929 Chr.19: 35250295-35250295 on Build GRCh38
rs769129288 Chr.19: 35250418-35250418 on Build GRCh38
rs781366796 Chr.19: 35250396-35250396 on Build GRCh38
rs370997772 Chr.19: 35250420-35250420 on Build GRCh38
rs747040466 Chr.19: 35250264-35250264 on Build GRCh38
rs747326286 Chr.19: 35250384-35250384 on Build GRCh38
rs776403442 Chr.19: 35250366-35250366 on Build GRCh38
rs564575842 Chr.19: 35250389-35250389 on Build GRCh38
rs763408450 Chr.19: 35250289-35250289 on Build GRCh38
rs141731488 Chr.19: 35250355-35250355 on Build GRCh38
rs371197377 Chr.19: 35250343-35250343 on Build GRCh38
rs1057463166 Chr.19: 35250273-35250273 on Build GRCh38
rs770438587 Chr.19: 35250282-35250282 on Build GRCh38
rs775949449 Chr.19: 35250288-35250288 on Build GRCh38
rs774582977 Chr.19: 35250419-35250419 on Build GRCh38
rs765660049 Chr.19: 35250401-35250402 on Build GRCh38
rs777846962 Chr.19: 35250372-35250372 on Build GRCh38
rs533002839 Chr.19: 35250359-35250359 on Build GRCh38
rs938733597 Chr.19: 35250260-35250260 on Build GRCh38
rs745967687 Chr.19: 35250405-35250405 on Build GRCh38
rs763624775 Chr.19: 35250293-35250293 on Build GRCh38
rs376231231 Chr.19: 35250301-35250301 on Build GRCh38
rs550867381 Chr.19: 35250255-35250255 on Build GRCh38
rs771067239 Chr.19: 35250391-35250391 on Build GRCh38
rs569450893 Chr.19: 35250416-35250416 on Build GRCh38
rs1044349389 Chr.19: 35250252-35250252 on Build GRCh38
rs766984646 Chr.19: 35250340-35250340 on Build GRCh38
rs1023367592 Chr.19: 35250361-35250361 on Build GRCh38
rs1053545044 Chr.19: 35250238-35250238 on Build GRCh38
rs868392352 Chr.19: 35250297-35250297 on Build GRCh38
rs771102831 Chr.19: 35250270-35250270 on Build GRCh38

View Full Product Details