Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 15177614-15177869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC39B
Entrez Gene ID
158219
Gene Name
tetratricopeptide repeat domain 39B
Gene Aliases
C9orf52
UniGene
Hs.563630
Gene Chromosome Location
Chr.9: 15167204-15307360 on Build GRCh38
Transcripts
NM_001168339.1 NM_001168340.1 NM_001168341.1 NM_001168342.1 NM_152574.2 XM_011517732.2 XM_017014310.1 XM_017014311.1 XM_017014312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6115279 c.1742G>T p.C581F Chr.9: 15177757-15177757 on Build GRCh38
6115276 c.1715G>A p.C572Y Chr.9: 15177784-15177784 on Build GRCh38
4152215 c.1536C>T p.S512S Chr.9: 15177804-15177804 on Build GRCh38
455696 c.1623T>C p.C541C Chr.9: 15177717-15177717 on Build GRCh38
1489865 c.1821T>C p.C607C Chr.9: 15177717-15177717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757968118 Chr.9: 15177719-15177719 on Build GRCh38
rs577613547 Chr.9: 15177807-15177807 on Build GRCh38
rs189065187 Chr.9: 15177801-15177801 on Build GRCh38
rs764948699 Chr.9: 15177740-15177740 on Build GRCh38
rs566023458 Chr.9: 15177674-15177674 on Build GRCh38
rs267602187 Chr.9: 15177755-15177755 on Build GRCh38
rs766512480 Chr.9: 15177773-15177773 on Build GRCh38
rs576608143 Chr.9: 15177739-15177739 on Build GRCh38
rs766205771 Chr.9: 15177685-15177685 on Build GRCh38
rs888941386 Chr.9: 15177706-15177706 on Build GRCh38
rs773006613 Chr.9: 15177822-15177822 on Build GRCh38
rs767600469 Chr.9: 15177676-15177676 on Build GRCh38
rs753712729 Chr.9: 15177696-15177696 on Build GRCh38
rs536235935 Chr.9: 15177683-15177683 on Build GRCh38
rs368846870 Chr.9: 15177832-15177844 on Build GRCh38
rs775035358 Chr.9: 15177744-15177744 on Build GRCh38
rs756222530 Chr.9: 15177648-15177648 on Build GRCh38
rs77494837 Chr.9: 15177720-15177720 on Build GRCh38
rs759378362 Chr.9: 15177792-15177792 on Build GRCh38
rs764682188 Chr.9: 15177710-15177710 on Build GRCh38
rs781039379 Chr.9: 15177829-15177829 on Build GRCh38
rs773246718 Chr.9: 15177760-15177760 on Build GRCh38
rs757834350 Chr.9: 15177809-15177809 on Build GRCh38
rs751131334 Chr.9: 15177822-15177822 on Build GRCh38
rs770260714 Chr.9: 15177667-15177667 on Build GRCh38
rs1000254264 Chr.9: 15177688-15177688 on Build GRCh38
rs114328012 Chr.9: 15177672-15177672 on Build GRCh38
rs780596213 Chr.9: 15177738-15177738 on Build GRCh38
rs200123094 Chr.9: 15177669-15177669 on Build GRCh38
rs756956909 Chr.9: 15177828-15177828 on Build GRCh38
rs763674446 Chr.9: 15177819-15177819 on Build GRCh38
rs753800978 Chr.9: 15177833-15177834 on Build GRCh38
rs752183843 Chr.9: 15177712-15177712 on Build GRCh38
rs746188407 Chr.9: 15177654-15177654 on Build GRCh38
rs756376183 Chr.9: 15177731-15177731 on Build GRCh38
rs766126168 Chr.9: 15177785-15177785 on Build GRCh38
rs760394157 Chr.9: 15177822-15177823 on Build GRCh38
rs762000975 Chr.9: 15177675-15177675 on Build GRCh38
rs890621698 Chr.9: 15177779-15177779 on Build GRCh38
rs754943659 Chr.9: 15177700-15177700 on Build GRCh38
rs370436765 Chr.9: 15177670-15177670 on Build GRCh38
rs746734967 Chr.9: 15177724-15177724 on Build GRCh38
rs750376501 Chr.9: 15177742-15177742 on Build GRCh38
rs148016922 Chr.9: 15177780-15177780 on Build GRCh38
rs377291867 Chr.9: 15177750-15177750 on Build GRCh38
rs971730674 Chr.9: 15177752-15177752 on Build GRCh38
rs267602188 Chr.9: 15177770-15177770 on Build GRCh38

View Full Product Details