Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11273218-11273489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRM3
Entrez Gene ID
58531
Gene Name
protamine 3
Gene Aliases
-
UniGene
Hs.259636
Gene Chromosome Location
Chr.16: 11273199-11273595 on Build GRCh38
Transcripts
NM_021247.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7225779 c.258C>G p.H86Q Chr.16: 11273338-11273338 on Build GRCh38
1375809 c.241C>T p.R81W Chr.16: 11273355-11273355 on Build GRCh38
7300444 c.207G>T p.P69P Chr.16: 11273389-11273389 on Build GRCh38
6538460 c.242G>C p.R81P Chr.16: 11273354-11273354 on Build GRCh38
6538461 c.166G>C p.E56Q Chr.16: 11273430-11273430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753493449 Chr.16: 11273345-11273345 on Build GRCh38
rs756977444 Chr.16: 11273450-11273450 on Build GRCh38
rs372340081 Chr.16: 11273458-11273458 on Build GRCh38
rs757587308 Chr.16: 11273348-11273348 on Build GRCh38
rs911408758 Chr.16: 11273405-11273405 on Build GRCh38
rs775588329 Chr.16: 11273258-11273258 on Build GRCh38
rs747088619 Chr.16: 11273271-11273271 on Build GRCh38
rs763652692 Chr.16: 11273446-11273446 on Build GRCh38
rs922846038 Chr.16: 11273423-11273440 on Build GRCh38
rs773402637 Chr.16: 11273404-11273405 on Build GRCh38
rs35598356 Chr.16: 11273338-11273338 on Build GRCh38
rs201908654 Chr.16: 11273390-11273390 on Build GRCh38
rs376051432 Chr.16: 11273389-11273389 on Build GRCh38
rs946282056 Chr.16: 11273447-11273447 on Build GRCh38
rs904410058 Chr.16: 11273431-11273431 on Build GRCh38
rs577224129 Chr.16: 11273387-11273387 on Build GRCh38
rs759245174 Chr.16: 11273423-11273431 on Build GRCh38
rs429744 Chr.16: 11273297-11273297 on Build GRCh38
rs556806318 Chr.16: 11273437-11273437 on Build GRCh38
rs894969575 Chr.16: 11273298-11273298 on Build GRCh38
rs759565179 Chr.16: 11273294-11273294 on Build GRCh38
rs1030449644 Chr.16: 11273276-11273276 on Build GRCh38
rs775506082 Chr.16: 11273443-11273443 on Build GRCh38
rs766156271 Chr.16: 11273289-11273289 on Build GRCh38
rs438289 Chr.16: 11273286-11273286 on Build GRCh38
rs948352368 Chr.16: 11273395-11273395 on Build GRCh38
rs529108983 Chr.16: 11273260-11273260 on Build GRCh38
rs1025730610 Chr.16: 11273373-11273373 on Build GRCh38
rs772438312 Chr.16: 11273247-11273247 on Build GRCh38
rs768841718 Chr.16: 11273253-11273254 on Build GRCh38
rs780526187 Chr.16: 11273376-11273376 on Build GRCh38
rs200105527 Chr.16: 11273445-11273445 on Build GRCh38
rs917015601 Chr.16: 11273393-11273393 on Build GRCh38
rs972461254 Chr.16: 11273361-11273361 on Build GRCh38
rs774175522 Chr.16: 11273291-11273291 on Build GRCh38
rs910771410 Chr.16: 11273309-11273309 on Build GRCh38
rs768924305 Chr.16: 11273280-11273280 on Build GRCh38
rs762341336 Chr.16: 11273389-11273390 on Build GRCh38
rs753268383 Chr.16: 11273449-11273449 on Build GRCh38
rs372747845 Chr.16: 11273326-11273326 on Build GRCh38
rs748507374 Chr.16: 11273384-11273384 on Build GRCh38
rs759244742 Chr.16: 11273418-11273418 on Build GRCh38
rs780277015 Chr.16: 11273281-11273281 on Build GRCh38
rs776775426 Chr.16: 11273283-11273283 on Build GRCh38
rs573666104 Chr.16: 11273457-11273457 on Build GRCh38
rs551217885 Chr.16: 11273325-11273325 on Build GRCh38
rs376082577 Chr.16: 11273329-11273329 on Build GRCh38
rs901190169 Chr.16: 11273241-11273241 on Build GRCh38
rs766957133 Chr.16: 11273457-11273457 on Build GRCh38
rs144080506 Chr.16: 11273275-11273275 on Build GRCh38
rs376400498 Chr.16: 11273355-11273355 on Build GRCh38
rs746340487 Chr.16: 11273243-11273243 on Build GRCh38
rs770061713 Chr.16: 11273407-11273407 on Build GRCh38
rs774239627 Chr.16: 11273410-11273410 on Build GRCh38
rs373206279 Chr.16: 11273354-11273354 on Build GRCh38
rs571306896 Chr.16: 11273377-11273378 on Build GRCh38
rs770366993 Chr.16: 11273404-11273421 on Build GRCh38
rs766697725 Chr.16: 11273418-11273420 on Build GRCh38
rs561407808 Chr.16: 11273242-11273242 on Build GRCh38
rs763357634 Chr.16: 11273415-11273415 on Build GRCh38
rs952517713 Chr.16: 11273381-11273381 on Build GRCh38
rs34977787 Chr.16: 11273360-11273360 on Build GRCh38
rs758703950 Chr.16: 11273460-11273460 on Build GRCh38
rs370987191 Chr.16: 11273386-11273386 on Build GRCh38
rs751209816 Chr.16: 11273422-11273423 on Build GRCh38
rs763847520 Chr.16: 11273343-11273343 on Build GRCh38

View Full Product Details