Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1815995-1816268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR1909
Entrez Gene ID
100302210
Gene Name
microRNA 1909
Gene Aliases
MIRN1909, hsa-mir-1909, mir-1909
UniGene
Gene Chromosome Location
Chr.19: 1816159-1816238 on Build GRCh38
Transcripts
NR_031730.1 NM_020695.3 XM_011528144.1 XM_011528146.2 XM_017027028.1 XM_017027029.1

Gene Symbol
REXO1
Entrez Gene ID
57455
Gene Name
RNA exonuclease 1 homolog
Gene Aliases
ELOABP1, EloA-BP1, REX1, TCEB3BP1
UniGene
Hs.192477
Gene Chromosome Location
Chr.19: 1815246-1848684 on Build GRCh38
Transcripts
NR_031730.1 NM_020695.3 XM_011528144.1 XM_011528146.2 XM_017027028.1 XM_017027029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291761 c.3578-4G>A p.? Chr.19: 1816158-1816158 on Build GRCh38
992979 c.3557G>T p.R1186I Chr.19: 1816245-1816245 on Build GRCh38
992978 c.3648C>T p.D1216D Chr.19: 1816084-1816084 on Build GRCh38
4825822 c.3603C>T p.D1201D Chr.19: 1816129-1816129 on Build GRCh38
4623872 c.3534G>A p.R1178R Chr.19: 1816268-1816268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562400060 Chr.19: 1816189-1816189 on Build GRCh38
rs548857767 Chr.19: 1816046-1816046 on Build GRCh38
rs779083064 Chr.19: 1816056-1816056 on Build GRCh38
rs780290937 Chr.19: 1816104-1816104 on Build GRCh38
rs376233777 Chr.19: 1816033-1816033 on Build GRCh38
rs567134814 Chr.19: 1816057-1816057 on Build GRCh38
rs369050526 Chr.19: 1816216-1816216 on Build GRCh38
rs773660112 Chr.19: 1816215-1816215 on Build GRCh38
rs34012959 Chr.19: 1816135-1816135 on Build GRCh38
rs748512305 Chr.19: 1816203-1816203 on Build GRCh38
rs768977406 Chr.19: 1816015-1816015 on Build GRCh38
rs370241664 Chr.19: 1816030-1816030 on Build GRCh38
rs765133979 Chr.19: 1816190-1816190 on Build GRCh38
rs758535327 Chr.19: 1816193-1816193 on Build GRCh38
rs763988557 Chr.19: 1816180-1816180 on Build GRCh38
rs771724752 Chr.19: 1816226-1816226 on Build GRCh38
rs928020021 Chr.19: 1816063-1816063 on Build GRCh38
rs747244196 Chr.19: 1816196-1816196 on Build GRCh38
rs753539930 Chr.19: 1816181-1816181 on Build GRCh38
rs917203730 Chr.19: 1816139-1816139 on Build GRCh38
rs777184637 Chr.19: 1816028-1816028 on Build GRCh38
rs781565881 Chr.19: 1816198-1816198 on Build GRCh38
rs368517238 Chr.19: 1816123-1816123 on Build GRCh38
rs138676883 Chr.19: 1816064-1816064 on Build GRCh38
rs79814521 Chr.19: 1816244-1816244 on Build GRCh38
rs890883731 Chr.19: 1816069-1816069 on Build GRCh38
rs769065488 Chr.19: 1816111-1816111 on Build GRCh38
rs376605843 Chr.19: 1816159-1816159 on Build GRCh38
rs201670065 Chr.19: 1816091-1816091 on Build GRCh38
rs750340588 Chr.19: 1816191-1816191 on Build GRCh38
rs774641337 Chr.19: 1816128-1816128 on Build GRCh38
rs770164595 Chr.19: 1816208-1816208 on Build GRCh38
rs199686292 Chr.19: 1816177-1816177 on Build GRCh38
rs200703160 Chr.19: 1816110-1816110 on Build GRCh38
rs901535136 Chr.19: 1816169-1816169 on Build GRCh38
rs777700881 Chr.19: 1816049-1816049 on Build GRCh38
rs1017785459 Chr.19: 1816085-1816085 on Build GRCh38
rs530762247 Chr.19: 1816029-1816029 on Build GRCh38
rs35839199 Chr.19: 1816016-1816017 on Build GRCh38
rs374156408 Chr.19: 1816158-1816158 on Build GRCh38
rs1044243564 Chr.19: 1816034-1816034 on Build GRCh38
rs372526700 Chr.19: 1816129-1816129 on Build GRCh38
rs760438108 Chr.19: 1816230-1816230 on Build GRCh38
rs775077981 Chr.19: 1816227-1816227 on Build GRCh38
rs199597990 Chr.19: 1816065-1816065 on Build GRCh38
rs375900640 Chr.19: 1816210-1816210 on Build GRCh38
rs767329272 Chr.19: 1816045-1816045 on Build GRCh38
rs1007017891 Chr.19: 1816071-1816071 on Build GRCh38
rs867554272 Chr.19: 1816232-1816232 on Build GRCh38
rs965219402 Chr.19: 1816122-1816122 on Build GRCh38
rs755125547 Chr.19: 1816197-1816197 on Build GRCh38
rs757078721 Chr.19: 1816053-1816053 on Build GRCh38
rs537748308 Chr.19: 1816170-1816170 on Build GRCh38
rs142716948 Chr.19: 1816084-1816084 on Build GRCh38
rs763793675 Chr.19: 1816243-1816243 on Build GRCh38
rs756040782 Chr.19: 1816047-1816047 on Build GRCh38
rs759256753 Chr.19: 1816043-1816043 on Build GRCh38
rs150797218 Chr.19: 1816125-1816125 on Build GRCh38

View Full Product Details