Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155803516-155803755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHH
Entrez Gene ID
6469
Gene Name
sonic hedgehog
Gene Aliases
HHG1, HLP3, HPE3, MCOPCB5, SMMCI, TPT, TPTPS
UniGene
Hs.164537
Gene Chromosome Location
Chr.7: 155799984-155812273 on Build GRCh38
Transcripts
NM_000193.3 NM_001310462.1 NR_132318.1 NR_132319.1 XM_011516479.2 XM_011516480.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6410299 c.578C>T p.A193V Chr.7: 155803711-155803711 on Build GRCh38
4763025 c.667C>T p.R223C Chr.7: 155803622-155803622 on Build GRCh38
485123 c.683A>G p.D228G Chr.7: 155803606-155803606 on Build GRCh38
1087863 c.732C>T p.R244R Chr.7: 155803557-155803557 on Build GRCh38
231147 c.619C>A p.L207M Chr.7: 155803670-155803670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780398556 Chr.7: 155803540-155803540 on Build GRCh38
rs771687521 Chr.7: 155803604-155803604 on Build GRCh38
rs779670631 Chr.7: 155803539-155803539 on Build GRCh38
rs374095917 Chr.7: 155803644-155803644 on Build GRCh38
rs999942157 Chr.7: 155803692-155803692 on Build GRCh38
rs896769031 Chr.7: 155803557-155803557 on Build GRCh38
rs553798627 Chr.7: 155803556-155803556 on Build GRCh38
rs587778803 Chr.7: 155803664-155803664 on Build GRCh38
rs199513347 Chr.7: 155803559-155803559 on Build GRCh38
rs754898783 Chr.7: 155803548-155803548 on Build GRCh38
rs587778806 Chr.7: 155803581-155803581 on Build GRCh38
rs756396169 Chr.7: 155803538-155803538 on Build GRCh38
rs766630931 Chr.7: 155803654-155803654 on Build GRCh38
rs9333633 Chr.7: 155803719-155803719 on Build GRCh38
rs754810872 Chr.7: 155803662-155803662 on Build GRCh38
rs774823152 Chr.7: 155803630-155803630 on Build GRCh38
rs1053198748 Chr.7: 155803678-155803678 on Build GRCh38
rs746540891 Chr.7: 155803709-155803709 on Build GRCh38
rs775658492 Chr.7: 155803620-155803620 on Build GRCh38
rs767551681 Chr.7: 155803646-155803646 on Build GRCh38
rs769075455 Chr.7: 155803628-155803628 on Build GRCh38
rs752650571 Chr.7: 155803702-155803702 on Build GRCh38
rs748157342 Chr.7: 155803585-155803585 on Build GRCh38
rs760754275 Chr.7: 155803653-155803653 on Build GRCh38
rs1012924397 Chr.7: 155803605-155803605 on Build GRCh38
rs104894043 Chr.7: 155803613-155803613 on Build GRCh38
rs368283830 Chr.7: 155803667-155803667 on Build GRCh38
rs746603362 Chr.7: 155803612-155803612 on Build GRCh38
rs777383609 Chr.7: 155803608-155803608 on Build GRCh38
rs780893190 Chr.7: 155803713-155803713 on Build GRCh38
rs372353493 Chr.7: 155803704-155803704 on Build GRCh38
rs104894042 Chr.7: 155803618-155803618 on Build GRCh38
rs763380033 Chr.7: 155803623-155803623 on Build GRCh38
rs750544127 Chr.7: 155803647-155803647 on Build GRCh38
rs9333634 Chr.7: 155803659-155803659 on Build GRCh38
rs587778805 Chr.7: 155803625-155803625 on Build GRCh38
rs948650219 Chr.7: 155803543-155803543 on Build GRCh38
rs770421409 Chr.7: 155803712-155803712 on Build GRCh38
rs768983299 Chr.7: 155803731-155803731 on Build GRCh38

View Full Product Details