Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30339779-30340052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM39
Entrez Gene ID
56658
Gene Name
tripartite motif containing 39
Gene Aliases
RNF23, TFP, TRIM39B
UniGene
Hs.413493
Gene Chromosome Location
Chr.6: 30326844-30343729 on Build GRCh38
Transcripts
NM_021253.3 NM_172016.2 NM_001199119.1

Gene Symbol
TRIM39-RPP21
Entrez Gene ID
202658
Gene Name
TRIM39-RPP21 readthrough
Gene Aliases
TRIM39R
UniGene
Hs.413493
Gene Chromosome Location
Chr.6: 30329311-30346858 on Build GRCh38
Transcripts
NM_021253.3 NM_172016.2 NM_001199119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5870036 c.799G>A p.E267K Chr.6: 30339926-30339926 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759519427 Chr.6: 30339857-30339857 on Build GRCh38
rs763824410 Chr.6: 30339911-30339911 on Build GRCh38
rs948616939 Chr.6: 30339841-30339841 on Build GRCh38
rs147338772 Chr.6: 30339970-30339970 on Build GRCh38
rs935061253 Chr.6: 30339879-30339879 on Build GRCh38
rs374065493 Chr.6: 30340003-30340003 on Build GRCh38
rs1006910113 Chr.6: 30340013-30340013 on Build GRCh38
rs761542230 Chr.6: 30339939-30339939 on Build GRCh38
rs535400224 Chr.6: 30339845-30339845 on Build GRCh38
rs749553323 Chr.6: 30339951-30339951 on Build GRCh38
rs768567364 Chr.6: 30339975-30339978 on Build GRCh38
rs781162919 Chr.6: 30339821-30339821 on Build GRCh38
rs762324428 Chr.6: 30339898-30339898 on Build GRCh38
rs890019462 Chr.6: 30340011-30340011 on Build GRCh38
rs568658122 Chr.6: 30339966-30339966 on Build GRCh38
rs775101590 Chr.6: 30339888-30339888 on Build GRCh38
rs749159502 Chr.6: 30340033-30340033 on Build GRCh38
rs781289955 Chr.6: 30340009-30340009 on Build GRCh38
rs769591912 Chr.6: 30339870-30339870 on Build GRCh38
rs775054944 Chr.6: 30339837-30339837 on Build GRCh38
rs200614440 Chr.6: 30339959-30339959 on Build GRCh38
rs1053698833 Chr.6: 30339894-30339894 on Build GRCh38
rs377153249 Chr.6: 30339960-30339960 on Build GRCh38
rs750781918 Chr.6: 30339942-30339942 on Build GRCh38
rs947532957 Chr.6: 30339999-30339999 on Build GRCh38
rs756720320 Chr.6: 30339950-30339950 on Build GRCh38
rs759227238 Chr.6: 30339915-30339915 on Build GRCh38
rs748429811 Chr.6: 30339982-30339982 on Build GRCh38
rs768039219 Chr.6: 30339941-30339941 on Build GRCh38

View Full Product Details