Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 94603341-94603574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASD1
Entrez Gene ID
64921
Gene Name
CAS1 domain containing 1
Gene Aliases
C7orf12, NBLA04196
UniGene
Hs.260041
Gene Chromosome Location
Chr.7: 94509858-94633465 on Build GRCh38
Transcripts
XM_011516495.2 NM_001099400.1 NM_001099401.1 NM_001301139.1 NM_003919.2 XM_005250675.4 XM_005250677.4 XM_011516663.2 XM_011516664.2 XM_011516665.2 XM_011516666.2 XM_011516667.2 XM_011516669.2 XM_017012763.1 XM_017012764.1 XM_017012765.1 XM_017012766.1 XM_017012767.1 XM_017012768.1 XM_017012769.1

Gene Symbol
SGCE
Entrez Gene ID
8910
Gene Name
sarcoglycan epsilon
Gene Aliases
DYT11, ESG
UniGene
Hs.371199
Gene Chromosome Location
Chr.7: 94585224-94656209 on Build GRCh38
Transcripts
XM_011516495.2 NM_001099400.1 NM_001099401.1 NM_001301139.1 NM_003919.2 XM_005250675.4 XM_005250677.4 XM_011516663.2 XM_011516664.2 XM_011516665.2 XM_011516666.2 XM_011516667.2 XM_011516669.2 XM_017012763.1 XM_017012764.1 XM_017012765.1 XM_017012766.1 XM_017012767.1 XM_017012768.1 XM_017012769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4725403 c.751G>T p.E251* Chr.7: 94603364-94603364 on Build GRCh38
1093289 c.774T>C p.C258C Chr.7: 94603341-94603341 on Build GRCh38
1150796 c.679G>A p.G227S Chr.7: 94603436-94603436 on Build GRCh38
1093290 c.693G>A p.P231P Chr.7: 94603422-94603422 on Build GRCh38
4725404 c.751G>T p.E251* Chr.7: 94603364-94603364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969428171 Chr.7: 94603522-94603522 on Build GRCh38
rs398123812 Chr.7: 94603406-94603406 on Build GRCh38
rs190993040 Chr.7: 94603535-94603535 on Build GRCh38
rs780709842 Chr.7: 94603474-94603489 on Build GRCh38
rs748327900 Chr.7: 94603457-94603457 on Build GRCh38
rs754956483 Chr.7: 94603451-94603451 on Build GRCh38
rs755996611 Chr.7: 94603423-94603423 on Build GRCh38
rs142833106 Chr.7: 94603399-94603399 on Build GRCh38
rs752351941 Chr.7: 94603379-94603379 on Build GRCh38
rs757096943 Chr.7: 94603456-94603457 on Build GRCh38
rs774652355 Chr.7: 94603494-94603494 on Build GRCh38
rs201158658 Chr.7: 94603426-94603426 on Build GRCh38
rs377151565 Chr.7: 94603472-94603472 on Build GRCh38
rs958186178 Chr.7: 94603411-94603411 on Build GRCh38
rs757206832 Chr.7: 94603422-94603422 on Build GRCh38
rs767137229 Chr.7: 94603377-94603377 on Build GRCh38
rs1011137135 Chr.7: 94603497-94603497 on Build GRCh38
rs770044091 Chr.7: 94603460-94603460 on Build GRCh38
rs998557057 Chr.7: 94603389-94603389 on Build GRCh38
rs781270494 Chr.7: 94603452-94603452 on Build GRCh38
rs760437933 Chr.7: 94603383-94603383 on Build GRCh38
rs1057519246 Chr.7: 94603448-94603448 on Build GRCh38
rs1038323242 Chr.7: 94603525-94603525 on Build GRCh38
rs374149816 Chr.7: 94603445-94603445 on Build GRCh38
rs549213726 Chr.7: 94603441-94603441 on Build GRCh38
rs746397599 Chr.7: 94603495-94603495 on Build GRCh38
rs777703229 Chr.7: 94603529-94603529 on Build GRCh38
rs200912914 Chr.7: 94603420-94603420 on Build GRCh38
rs749670022 Chr.7: 94603486-94603486 on Build GRCh38
rs771460445 Chr.7: 94603490-94603490 on Build GRCh38
rs1024282207 Chr.7: 94603515-94603515 on Build GRCh38
rs200035109 Chr.7: 94603403-94603403 on Build GRCh38

View Full Product Details