Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 89786500-89786743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIPK2
Entrez Gene ID
8767
Gene Name
receptor interacting serine/threonine kinase 2
Gene Aliases
CARD3, CARDIAK, CCK, GIG30, RICK, RIP2
UniGene
Hs.103755
Gene Chromosome Location
Chr.8: 89757747-89791064 on Build GRCh38
Transcripts
NM_003821.5 XM_005251092.3 XM_011517357.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4993158 c.1039G>A p.G347R Chr.8: 89786602-89786602 on Build GRCh38
7387617 c.1066G>T p.G356C Chr.8: 89786629-89786629 on Build GRCh38
5617777 c.1121C>G p.S374C Chr.8: 89786684-89786684 on Build GRCh38
1330879 c.1072C>G p.P358A Chr.8: 89786635-89786635 on Build GRCh38
6013210 c.1043C>T p.S348F Chr.8: 89786606-89786606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747586469 Chr.8: 89786608-89786608 on Build GRCh38
rs980774523 Chr.8: 89786675-89786675 on Build GRCh38
rs747004908 Chr.8: 89786587-89786587 on Build GRCh38
rs367674479 Chr.8: 89786548-89786548 on Build GRCh38
rs200826927 Chr.8: 89786651-89786651 on Build GRCh38
rs201478995 Chr.8: 89786634-89786634 on Build GRCh38
rs371304420 Chr.8: 89786682-89786682 on Build GRCh38
rs779979791 Chr.8: 89786551-89786551 on Build GRCh38
rs1009833909 Chr.8: 89786531-89786531 on Build GRCh38
rs772697211 Chr.8: 89786613-89786613 on Build GRCh38
rs199570893 Chr.8: 89786646-89786646 on Build GRCh38
rs142087655 Chr.8: 89786692-89786692 on Build GRCh38
rs993040441 Chr.8: 89786609-89786609 on Build GRCh38
rs759961470 Chr.8: 89786625-89786625 on Build GRCh38
rs757244288 Chr.8: 89786672-89786672 on Build GRCh38
rs971247899 Chr.8: 89786552-89786552 on Build GRCh38
rs372772811 Chr.8: 89786696-89786696 on Build GRCh38
rs200625019 Chr.8: 89786600-89786600 on Build GRCh38
rs778955719 Chr.8: 89786545-89786545 on Build GRCh38
rs200218081 Chr.8: 89786689-89786689 on Build GRCh38
rs768228896 Chr.8: 89786633-89786635 on Build GRCh38
rs772207077 Chr.8: 89786550-89786550 on Build GRCh38
rs753995384 Chr.8: 89786670-89786670 on Build GRCh38
rs1022935870 Chr.8: 89786580-89786580 on Build GRCh38
rs769505144 Chr.8: 89786611-89786611 on Build GRCh38
rs764677295 Chr.8: 89786659-89786659 on Build GRCh38
rs199844994 Chr.8: 89786590-89786590 on Build GRCh38
rs754954132 Chr.8: 89786704-89786704 on Build GRCh38
rs776156245 Chr.8: 89786602-89786602 on Build GRCh38
rs745706979 Chr.8: 89786547-89786547 on Build GRCh38
rs36069506 Chr.8: 89786699-89786700 on Build GRCh38
rs765502397 Chr.8: 89786674-89786674 on Build GRCh38
rs961265424 Chr.8: 89786562-89786562 on Build GRCh38
rs201250830 Chr.8: 89786569-89786569 on Build GRCh38

View Full Product Details