Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45458154-45458415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHM1
Entrez Gene ID
9842
Gene Name
pleckstrin homology and RUN domain containing M1
Gene Aliases
AP162, B2, OPTB6
UniGene
Hs.514242
Gene Chromosome Location
Chr.17: 45434262-45490780 on Build GRCh38
Transcripts
NM_014798.2 NR_027774.1 NR_027782.1 XM_006722201.3 XM_011525523.2 XM_011525524.1 XM_011525525.1 XM_011525526.1 XM_017025451.1 XM_017025452.1 XM_017025453.1 XM_017025454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3518289 c.1349C>T p.P450L Chr.17: 45458399-45458399 on Build GRCh38
980162 c.1458A>G p.R486R Chr.17: 45458290-45458290 on Build GRCh38
4388520 c.1484A>C p.D495A Chr.17: 45458264-45458264 on Build GRCh38
980161 c.1478C>T p.A493V Chr.17: 45458270-45458270 on Build GRCh38
3518288 c.1397C>T p.S466F Chr.17: 45458351-45458351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147190961 Chr.17: 45458279-45458279 on Build GRCh38
rs755729074 Chr.17: 45458219-45458219 on Build GRCh38
rs774636888 Chr.17: 45458387-45458387 on Build GRCh38
rs747512253 Chr.17: 45458347-45458347 on Build GRCh38
rs550003692 Chr.17: 45458253-45458253 on Build GRCh38
rs770017312 Chr.17: 45458363-45458363 on Build GRCh38
rs756519411 Chr.17: 45458330-45458330 on Build GRCh38
rs779561311 Chr.17: 45458243-45458243 on Build GRCh38
rs923451297 Chr.17: 45458280-45458280 on Build GRCh38
rs757083828 Chr.17: 45458192-45458192 on Build GRCh38
rs2868616 Chr.17: 45458176-45458176 on Build GRCh38
rs140558219 Chr.17: 45458358-45458358 on Build GRCh38
rs561587976 Chr.17: 45458357-45458357 on Build GRCh38
rs554125896 Chr.17: 45458325-45458325 on Build GRCh38
rs769919860 Chr.17: 45458270-45458270 on Build GRCh38
rs753568143 Chr.17: 45458328-45458328 on Build GRCh38
rs761897823 Chr.17: 45458282-45458282 on Build GRCh38
rs937388027 Chr.17: 45458386-45458386 on Build GRCh38
rs964868552 Chr.17: 45458271-45458271 on Build GRCh38
rs761787525 Chr.17: 45458393-45458393 on Build GRCh38
rs765353297 Chr.17: 45458180-45458180 on Build GRCh38
rs764305349 Chr.17: 45458278-45458278 on Build GRCh38
rs371968983 Chr.17: 45458269-45458269 on Build GRCh38
rs752722549 Chr.17: 45458181-45458181 on Build GRCh38
rs1037801172 Chr.17: 45458384-45458384 on Build GRCh38
rs767278755 Chr.17: 45458306-45458306 on Build GRCh38
rs896415203 Chr.17: 45458216-45458216 on Build GRCh38
rs764149827 Chr.17: 45458188-45458188 on Build GRCh38
rs558775468 Chr.17: 45458326-45458326 on Build GRCh38
rs1013498579 Chr.17: 45458221-45458221 on Build GRCh38
rs531873798 Chr.17: 45458186-45458186 on Build GRCh38
rs781178979 Chr.17: 45458193-45458193 on Build GRCh38
rs772531601 Chr.17: 45458255-45458255 on Build GRCh38
rs750074826 Chr.17: 45458212-45458212 on Build GRCh38
rs778221542 Chr.17: 45458257-45458257 on Build GRCh38
rs767381421 Chr.17: 45458394-45458394 on Build GRCh38
rs757764009 Chr.17: 45458348-45458348 on Build GRCh38
rs775539977 Chr.17: 45458366-45458366 on Build GRCh38
rs1055394825 Chr.17: 45458390-45458390 on Build GRCh38
rs754463825 Chr.17: 45458336-45458336 on Build GRCh38
rs750626603 Chr.17: 45458200-45458200 on Build GRCh38
rs140977155 Chr.17: 45458385-45458385 on Build GRCh38
rs368968382 Chr.17: 45458258-45458258 on Build GRCh38
rs1021082250 Chr.17: 45458229-45458229 on Build GRCh38
rs778454341 Chr.17: 45458346-45458346 on Build GRCh38
rs571674138 Chr.17: 45458273-45458273 on Build GRCh38
rs749566636 Chr.17: 45458374-45458374 on Build GRCh38
rs751356371 Chr.17: 45458191-45458191 on Build GRCh38

View Full Product Details