Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24789595-24789820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF17
Entrez Gene ID
56163
Gene Name
ring finger protein 17
Gene Aliases
Mmip-2, SPATA23, TDRD4
UniGene
Hs.572245
Gene Chromosome Location
Chr.13: 24747641-24888816 on Build GRCh38
Transcripts
NM_001184993.1 NM_031277.2 XM_006719846.3 XM_006719849.2 XM_006719852.2 XM_011535152.2 XM_011535155.2 XM_011535156.2 XM_011535157.2 XM_011535158.2 XM_011535159.2 XM_011535160.2 XM_011535162.1 XM_017020676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3399285 c.900C>A p.N300K Chr.13: 24789737-24789737 on Build GRCh38
3700393 c.864G>T p.L288F Chr.13: 24789701-24789701 on Build GRCh38
7271338 c.935+6T>C p.? Chr.13: 24789778-24789778 on Build GRCh38
164055 c.885C>T p.I295I Chr.13: 24789722-24789722 on Build GRCh38
7271340 c.935+6T>C p.? Chr.13: 24789778-24789778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201724014 Chr.13: 24789764-24789764 on Build GRCh38
rs113784879 Chr.13: 24789650-24789650 on Build GRCh38
rs755205067 Chr.13: 24789784-24789784 on Build GRCh38
rs760972665 Chr.13: 24789776-24789776 on Build GRCh38
rs769722259 Chr.13: 24789720-24789720 on Build GRCh38
rs372404223 Chr.13: 24789779-24789779 on Build GRCh38
rs779025138 Chr.13: 24789789-24789789 on Build GRCh38
rs775965201 Chr.13: 24789703-24789703 on Build GRCh38
rs754073791 Chr.13: 24789662-24789662 on Build GRCh38
rs564854281 Chr.13: 24789632-24789632 on Build GRCh38
rs780185608 Chr.13: 24789658-24789658 on Build GRCh38
rs369578539 Chr.13: 24789769-24789769 on Build GRCh38
rs761251799 Chr.13: 24789765-24789765 on Build GRCh38
rs765644549 Chr.13: 24789747-24789747 on Build GRCh38
rs745589005 Chr.13: 24789715-24789715 on Build GRCh38
rs752778322 Chr.13: 24789793-24789793 on Build GRCh38
rs761426824 Chr.13: 24789739-24789739 on Build GRCh38
rs755220371 Chr.13: 24789663-24789663 on Build GRCh38
rs6490979 Chr.13: 24789638-24789638 on Build GRCh38
rs754239136 Chr.13: 24789783-24789783 on Build GRCh38
rs770849190 Chr.13: 24789694-24789694 on Build GRCh38
rs376276811 Chr.13: 24789729-24789729 on Build GRCh38
rs750737460 Chr.13: 24789652-24789652 on Build GRCh38
rs773769662 Chr.13: 24789763-24789763 on Build GRCh38
rs1005718108 Chr.13: 24789682-24789682 on Build GRCh38
rs1015180786 Chr.13: 24789719-24789719 on Build GRCh38
rs547483711 Chr.13: 24789678-24789678 on Build GRCh38
rs748224275 Chr.13: 24789693-24789693 on Build GRCh38
rs747127259 Chr.13: 24789771-24789774 on Build GRCh38
rs756503158 Chr.13: 24789653-24789653 on Build GRCh38

View Full Product Details