Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 141973576-141973801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAS2R38
Entrez Gene ID
5726
Gene Name
taste 2 receptor member 38
Gene Aliases
PTC, T2R38, T2R61
UniGene
Hs.647085
Gene Chromosome Location
Chr.7: 141972631-141973773 on Build GRCh38
Transcripts
NM_176817.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4396720 c.67G>A p.V23I Chr.7: 141973623-141973623 on Build GRCh38
2861737 c.105C>T p.F35F Chr.7: 141973585-141973585 on Build GRCh38
5639388 c.106G>A p.V36I Chr.7: 141973584-141973584 on Build GRCh38
5062247 c.88T>G p.F30V Chr.7: 141973602-141973602 on Build GRCh38
5872022 c.65C>T p.S22L Chr.7: 141973625-141973625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782429724 Chr.7: 141973721-141973721 on Build GRCh38
rs371843478 Chr.7: 141973782-141973782 on Build GRCh38
rs782033376 Chr.7: 141973702-141973702 on Build GRCh38
rs782730686 Chr.7: 141973690-141973690 on Build GRCh38
rs200360674 Chr.7: 141973705-141973705 on Build GRCh38
rs368901923 Chr.7: 141973662-141973662 on Build GRCh38
rs375058241 Chr.7: 141973674-141973674 on Build GRCh38
rs782176127 Chr.7: 141973717-141973717 on Build GRCh38
rs377342463 Chr.7: 141973668-141973668 on Build GRCh38
rs375588043 Chr.7: 141973667-141973667 on Build GRCh38
rs963782056 Chr.7: 141973748-141973748 on Build GRCh38
rs782149941 Chr.7: 141973774-141973774 on Build GRCh38
rs368123866 Chr.7: 141973706-141973706 on Build GRCh38
rs150525684 Chr.7: 141973683-141973683 on Build GRCh38
rs781811462 Chr.7: 141973671-141973671 on Build GRCh38
rs1019811396 Chr.7: 141973736-141973736 on Build GRCh38
rs781965322 Chr.7: 141973604-141973604 on Build GRCh38
rs781912967 Chr.7: 141973655-141973655 on Build GRCh38
rs541691931 Chr.7: 141973682-141973682 on Build GRCh38
rs370330094 Chr.7: 141973669-141973669 on Build GRCh38
rs372430424 Chr.7: 141973716-141973716 on Build GRCh38
rs975188101 Chr.7: 141973766-141973766 on Build GRCh38
rs782002473 Chr.7: 141973723-141973723 on Build GRCh38
rs556042962 Chr.7: 141973648-141973648 on Build GRCh38
rs782509604 Chr.7: 141973688-141973688 on Build GRCh38
rs782469789 Chr.7: 141973708-141973708 on Build GRCh38
rs371175533 Chr.7: 141973635-141973635 on Build GRCh38
rs782509053 Chr.7: 141973678-141973679 on Build GRCh38
rs141743784 Chr.7: 141973673-141973673 on Build GRCh38

View Full Product Details