Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6467957-6468224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHG5
Entrez Gene ID
57449
Gene Name
pleckstrin homology and RhoGEF domain containing G5
Gene Aliases
CMTRIC, DSMA4, GEF720, Syx, Tech
UniGene
Hs.284232
Gene Chromosome Location
Chr.1: 6466092-6520061 on Build GRCh38
Transcripts
NM_001042663.1 NM_001042664.1 NM_001042665.1 NM_001265592.1 NM_001265593.1 NM_001265594.1 NM_020631.4 NM_198681.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4642023 c.2873G>A p.S958N Chr.1: 6468194-6468194 on Build GRCh38
6342397 c.2897C>A p.A966D Chr.1: 6468170-6468170 on Build GRCh38
6383611 c.3083C>T p.P1028L Chr.1: 6467990-6467990 on Build GRCh38
4869887 c.2944G>T p.G982C Chr.1: 6468099-6468099 on Build GRCh38
4642024 c.2879G>A p.S960N Chr.1: 6468194-6468194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374594401 Chr.1: 6468151-6468151 on Build GRCh38
rs760436735 Chr.1: 6467994-6467994 on Build GRCh38
rs776844267 Chr.1: 6467977-6467977 on Build GRCh38
rs879254222 Chr.1: 6467988-6467988 on Build GRCh38
rs753662166 Chr.1: 6468153-6468153 on Build GRCh38
rs781226672 Chr.1: 6468167-6468167 on Build GRCh38
rs779825659 Chr.1: 6468037-6468037 on Build GRCh38
rs114619322 Chr.1: 6468009-6468009 on Build GRCh38
rs755699992 Chr.1: 6468018-6468018 on Build GRCh38
rs758308121 Chr.1: 6468160-6468160 on Build GRCh38
rs887770274 Chr.1: 6467994-6467994 on Build GRCh38
rs778854412 Chr.1: 6468048-6468048 on Build GRCh38
rs775809877 Chr.1: 6468103-6468103 on Build GRCh38
rs764378556 Chr.1: 6468116-6468116 on Build GRCh38
rs776959400 Chr.1: 6468089-6468089 on Build GRCh38
rs769702301 Chr.1: 6468194-6468194 on Build GRCh38
rs573883242 Chr.1: 6468024-6468024 on Build GRCh38
rs367560509 Chr.1: 6468202-6468202 on Build GRCh38
rs200407689 Chr.1: 6468159-6468159 on Build GRCh38
rs376237905 Chr.1: 6468047-6468047 on Build GRCh38
rs774696513 Chr.1: 6468123-6468123 on Build GRCh38
rs187886272 Chr.1: 6468090-6468090 on Build GRCh38
rs879144584 Chr.1: 6468093-6468093 on Build GRCh38
rs376406302 Chr.1: 6468155-6468155 on Build GRCh38
rs1050889710 Chr.1: 6467993-6467993 on Build GRCh38
rs750132016 Chr.1: 6468138-6468138 on Build GRCh38
rs748020400 Chr.1: 6468049-6468049 on Build GRCh38
rs370666430 Chr.1: 6468085-6468085 on Build GRCh38
rs763526966 Chr.1: 6468111-6468111 on Build GRCh38
rs886042289 Chr.1: 6468034-6468034 on Build GRCh38
rs765636442 Chr.1: 6467990-6467990 on Build GRCh38
rs910647086 Chr.1: 6468087-6468087 on Build GRCh38
rs756909775 Chr.1: 6468005-6468005 on Build GRCh38
rs573561537 Chr.1: 6467998-6467998 on Build GRCh38
rs749438508 Chr.1: 6468201-6468201 on Build GRCh38
rs527750670 Chr.1: 6468149-6468149 on Build GRCh38
rs746121801 Chr.1: 6468173-6468173 on Build GRCh38
rs762228019 Chr.1: 6468127-6468127 on Build GRCh38
rs374340207 Chr.1: 6468195-6468195 on Build GRCh38
rs755539639 Chr.1: 6468145-6468145 on Build GRCh38
rs749891873 Chr.1: 6468010-6468010 on Build GRCh38
rs767138737 Chr.1: 6468137-6468137 on Build GRCh38
rs751158651 Chr.1: 6467999-6467999 on Build GRCh38
rs779048924 Chr.1: 6468158-6468158 on Build GRCh38
rs759718802 Chr.1: 6467980-6467980 on Build GRCh38
rs753283144 Chr.1: 6467995-6467995 on Build GRCh38
rs555305602 Chr.1: 6467997-6467997 on Build GRCh38
rs536097668 Chr.1: 6468075-6468077 on Build GRCh38

View Full Product Details