Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6012948-6013179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP6
Entrez Gene ID
26953
Gene Name
RAN binding protein 6
Gene Aliases
-
UniGene
Hs.167496
Gene Chromosome Location
Chr.9: 6011019-6015660 on Build GRCh38
Transcripts
NM_001243202.1 NM_001243203.1 NM_012416.3 XM_017014624.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5902928 c.2456G>A p.R819K Chr.9: 6013152-6013152 on Build GRCh38
4163944 c.2592G>A p.K864K Chr.9: 6013016-6013016 on Build GRCh38
3907588 c.2504C>G p.S835C Chr.9: 6013104-6013104 on Build GRCh38
1176946 c.2483A>G p.Y828C Chr.9: 6013125-6013125 on Build GRCh38
6183607 c.2572T>C p.F858L Chr.9: 6013036-6013036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763111978 Chr.9: 6012989-6012989 on Build GRCh38
rs187892114 Chr.9: 6013008-6013008 on Build GRCh38
rs747257765 Chr.9: 6013115-6013115 on Build GRCh38
rs753811575 Chr.9: 6013091-6013091 on Build GRCh38
rs749835001 Chr.9: 6013058-6013058 on Build GRCh38
rs747204599 Chr.9: 6012974-6012974 on Build GRCh38
rs757518200 Chr.9: 6013135-6013135 on Build GRCh38
rs776524425 Chr.9: 6012998-6012998 on Build GRCh38
rs745352691 Chr.9: 6013137-6013137 on Build GRCh38
rs994549009 Chr.9: 6013030-6013030 on Build GRCh38
rs976544886 Chr.9: 6013123-6013123 on Build GRCh38
rs771184905 Chr.9: 6012995-6012995 on Build GRCh38
rs760124713 Chr.9: 6013006-6013006 on Build GRCh38
rs745882029 Chr.9: 6013146-6013146 on Build GRCh38
rs769662226 Chr.9: 6013037-6013037 on Build GRCh38
rs764408824 Chr.9: 6013029-6013029 on Build GRCh38
rs375551495 Chr.9: 6012986-6012986 on Build GRCh38
rs1411949 Chr.9: 6012968-6012968 on Build GRCh38
rs574224935 Chr.9: 6013027-6013027 on Build GRCh38
rs778829981 Chr.9: 6013108-6013108 on Build GRCh38
rs770135556 Chr.9: 6013007-6013007 on Build GRCh38
rs767084856 Chr.9: 6013054-6013054 on Build GRCh38
rs754862076 Chr.9: 6013097-6013097 on Build GRCh38
rs1001634296 Chr.9: 6013009-6013009 on Build GRCh38
rs201088423 Chr.9: 6013102-6013102 on Build GRCh38
rs139005279 Chr.9: 6013051-6013051 on Build GRCh38
rs368525268 Chr.9: 6013025-6013025 on Build GRCh38
rs779911838 Chr.9: 6013063-6013063 on Build GRCh38
rs756089965 Chr.9: 6013062-6013062 on Build GRCh38
rs751258787 Chr.9: 6013040-6013040 on Build GRCh38

View Full Product Details