Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37574377-37574638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POSTN
Entrez Gene ID
10631
Gene Name
periostin
Gene Aliases
OSF-2, OSF2, PDLPOSTN, PN
UniGene
Hs.136348
Gene Chromosome Location
Chr.13: 37562582-37598844 on Build GRCh38
Transcripts
NM_001135934.1 NM_001135935.1 NM_001135936.1 NM_001286665.1 NM_001286666.1 NM_001286667.1 NM_006475.2 XM_005266231.2 XM_005266232.2 XM_017020355.1 XM_017020356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4047450 c.2052G>A p.V684V Chr.13: 37574609-37574609 on Build GRCh38
7319155 c.2035G>T p.E679* Chr.13: 37574626-37574626 on Build GRCh38
1607056 c.2089+3A>T p.? Chr.13: 37574569-37574569 on Build GRCh38
220610 c.2050G>A p.V684M Chr.13: 37574611-37574611 on Build GRCh38
5342330 c.2089G>A p.G697R Chr.13: 37574572-37574572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991548614 Chr.13: 37574423-37574423 on Build GRCh38
rs575553035 Chr.13: 37574455-37574455 on Build GRCh38
rs202148360 Chr.13: 37574560-37574560 on Build GRCh38
rs745957279 Chr.13: 37574530-37574530 on Build GRCh38
rs369576714 Chr.13: 37574585-37574585 on Build GRCh38
rs924673990 Chr.13: 37574529-37574529 on Build GRCh38
rs753661205 Chr.13: 37574442-37574442 on Build GRCh38
rs922796487 Chr.13: 37574446-37574446 on Build GRCh38
rs765935819 Chr.13: 37574559-37574559 on Build GRCh38
rs759152372 Chr.13: 37574563-37574563 on Build GRCh38
rs573234513 Chr.13: 37574577-37574577 on Build GRCh38
rs1041286839 Chr.13: 37574541-37574541 on Build GRCh38
rs964296485 Chr.13: 37574439-37574439 on Build GRCh38
rs879794876 Chr.13: 37574478-37574478 on Build GRCh38
rs75996921 Chr.13: 37574549-37574549 on Build GRCh38
rs752458898 Chr.13: 37574587-37574587 on Build GRCh38
rs769926643 Chr.13: 37574532-37574532 on Build GRCh38
rs909931185 Chr.13: 37574516-37574516 on Build GRCh38
rs982862003 Chr.13: 37574492-37574492 on Build GRCh38
rs777855935 Chr.13: 37574601-37574601 on Build GRCh38
rs749529416 Chr.13: 37574545-37574545 on Build GRCh38
rs762194957 Chr.13: 37574550-37574550 on Build GRCh38
rs373246380 Chr.13: 37574548-37574548 on Build GRCh38
rs558785615 Chr.13: 37574431-37574431 on Build GRCh38
rs930131136 Chr.13: 37574449-37574449 on Build GRCh38
rs372244963 Chr.13: 37574588-37574588 on Build GRCh38
rs772586596 Chr.13: 37574552-37574552 on Build GRCh38
rs554598370 Chr.13: 37574558-37574558 on Build GRCh38
rs947521933 Chr.13: 37574438-37574438 on Build GRCh38
rs181222797 Chr.13: 37574416-37574416 on Build GRCh38
rs777805116 Chr.13: 37574527-37574527 on Build GRCh38
rs780276994 Chr.13: 37574539-37574539 on Build GRCh38
rs768897966 Chr.13: 37574546-37574546 on Build GRCh38
rs1043441987 Chr.13: 37574456-37574456 on Build GRCh38
rs976907066 Chr.13: 37574440-37574440 on Build GRCh38

View Full Product Details