Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 61045263-61045502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A6
Entrez Gene ID
145389
Gene Name
solute carrier family 38 member 6
Gene Aliases
NAT-1, SNAT6
UniGene
Hs.200738
Gene Chromosome Location
Chr.14: 60981114-61083733 on Build GRCh38
Transcripts
NM_001172702.1 NM_153811.2 NR_033344.1 XM_006720049.3 XM_006720050.1 XM_011536469.2 XM_017021020.1 XM_017021021.1 XM_017021022.1 XM_017021023.1 XM_017021024.1 XM_017021025.1 XM_017021026.1 XM_017021027.1 XM_017021028.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6203686 c.754G>A p.A252T Chr.14: 61045355-61045355 on Build GRCh38
470092 c.761C>T p.P254L Chr.14: 61045362-61045362 on Build GRCh38
6335994 c.814G>T p.E272* Chr.14: 61045415-61045415 on Build GRCh38
6203687 c.754G>A p.A252T Chr.14: 61045355-61045355 on Build GRCh38
6554438 c.809A>T p.Y270F Chr.14: 61045410-61045410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757870634 Chr.14: 61045430-61045430 on Build GRCh38
rs45523838 Chr.14: 61045353-61045353 on Build GRCh38
rs779844978 Chr.14: 61045406-61045406 on Build GRCh38
rs906904886 Chr.14: 61045433-61045433 on Build GRCh38
rs776428829 Chr.14: 61045437-61045437 on Build GRCh38
rs762729186 Chr.14: 61045469-61045469 on Build GRCh38
rs757266919 Chr.14: 61045364-61045364 on Build GRCh38
rs868414334 Chr.14: 61045377-61045377 on Build GRCh38
rs144811998 Chr.14: 61045418-61045418 on Build GRCh38
rs762904896 Chr.14: 61045410-61045411 on Build GRCh38
rs769444729 Chr.14: 61045459-61045459 on Build GRCh38
rs768679296 Chr.14: 61045428-61045428 on Build GRCh38
rs756670398 Chr.14: 61045344-61045344 on Build GRCh38
rs762639254 Chr.14: 61045311-61045311 on Build GRCh38
rs923882838 Chr.14: 61045329-61045329 on Build GRCh38
rs768000534 Chr.14: 61045327-61045327 on Build GRCh38
rs772240122 Chr.14: 61045397-61045397 on Build GRCh38
rs778903962 Chr.14: 61045370-61045370 on Build GRCh38
rs1056310533 Chr.14: 61045400-61045400 on Build GRCh38
rs755758782 Chr.14: 61045378-61045378 on Build GRCh38
rs374253278 Chr.14: 61045297-61045297 on Build GRCh38
rs752967634 Chr.14: 61045328-61045328 on Build GRCh38
rs1013802480 Chr.14: 61045411-61045411 on Build GRCh38
rs746093089 Chr.14: 61045387-61045387 on Build GRCh38
rs931512680 Chr.14: 61045329-61045330 on Build GRCh38
rs547137539 Chr.14: 61045350-61045350 on Build GRCh38
rs772783814 Chr.14: 61045464-61045464 on Build GRCh38
rs750088693 Chr.14: 61045304-61045304 on Build GRCh38
rs761445244 Chr.14: 61045457-61045457 on Build GRCh38
rs370602836 Chr.14: 61045367-61045367 on Build GRCh38

View Full Product Details