Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58351323-58351596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT4H1
Entrez Gene ID
6827
Gene Name
SPT4 homolog, DSIF elongation factor subunit
Gene Aliases
SPT4, SPT4H, SUPT4H, Supt4a
UniGene
Hs.439481
Gene Chromosome Location
Chr.17: 58345175-58352238 on Build GRCh38
Transcripts
NM_003168.2 NR_073470.1 NR_038410.1 NR_038411.1 NR_038412.1 NR_038413.1 NR_038414.1 NR_038415.1 NR_038416.1 NR_038417.1 NR_038418.1

Gene Symbol
TSPOAP1-AS1
Entrez Gene ID
100506779
Gene Name
TSPOAP1 antisense RNA 1
Gene Aliases
BZRAP1-AS1
UniGene
Hs.718477
Gene Chromosome Location
Chr.17: 58325450-58353727 on Build GRCh38
Transcripts
NM_003168.2 NR_073470.1 NR_038410.1 NR_038411.1 NR_038412.1 NR_038413.1 NR_038414.1 NR_038415.1 NR_038416.1 NR_038417.1 NR_038418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6556518 c.76G>A p.D26N Chr.17: 58351502-58351502 on Build GRCh38
1303110 c.139G>A p.E47K Chr.17: 58351439-58351439 on Build GRCh38
6046374 c.103A>T p.N35Y Chr.17: 58351475-58351475 on Build GRCh38
5799965 c.70-4C>A p.? Chr.17: 58351512-58351512 on Build GRCh38
7321572 c.105T>A p.N35K Chr.17: 58351473-58351473 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772670710 Chr.17: 58351377-58351379 on Build GRCh38
rs768095539 Chr.17: 58351395-58351395 on Build GRCh38
rs758073287 Chr.17: 58351374-58351374 on Build GRCh38
rs747709634 Chr.17: 58351547-58351547 on Build GRCh38
rs767553011 Chr.17: 58351468-58351468 on Build GRCh38
rs147770689 Chr.17: 58351367-58351367 on Build GRCh38
rs753194461 Chr.17: 58351357-58351357 on Build GRCh38
rs779880233 Chr.17: 58351376-58351376 on Build GRCh38
rs774545118 Chr.17: 58351566-58351566 on Build GRCh38
rs201993962 Chr.17: 58351513-58351513 on Build GRCh38
rs746483377 Chr.17: 58351380-58351380 on Build GRCh38
rs749051696 Chr.17: 58351564-58351564 on Build GRCh38
rs762762508 Chr.17: 58351449-58351449 on Build GRCh38
rs922114806 Chr.17: 58351460-58351460 on Build GRCh38
rs371112173 Chr.17: 58351411-58351411 on Build GRCh38
rs761028956 Chr.17: 58351473-58351473 on Build GRCh38
rs374457896 Chr.17: 58351351-58351351 on Build GRCh38
rs527644760 Chr.17: 58351359-58351359 on Build GRCh38
rs535652912 Chr.17: 58351560-58351560 on Build GRCh38
rs759638967 Chr.17: 58351465-58351465 on Build GRCh38
rs778216581 Chr.17: 58351366-58351366 on Build GRCh38
rs764718186 Chr.17: 58351504-58351504 on Build GRCh38
rs751225358 Chr.17: 58351532-58351532 on Build GRCh38
rs769779914 Chr.17: 58351442-58351442 on Build GRCh38
rs754214027 Chr.17: 58351510-58351510 on Build GRCh38
rs748103739 Chr.17: 58351420-58351420 on Build GRCh38
rs769673560 Chr.17: 58351556-58351556 on Build GRCh38
rs774598303 Chr.17: 58351464-58351464 on Build GRCh38
rs759745059 Chr.17: 58351571-58351571 on Build GRCh38
rs141480962 Chr.17: 58351443-58351443 on Build GRCh38
rs568124021 Chr.17: 58351544-58351544 on Build GRCh38
rs942453172 Chr.17: 58351429-58351429 on Build GRCh38
rs375632531 Chr.17: 58351554-58351554 on Build GRCh38
rs147819416 Chr.17: 58351470-58351470 on Build GRCh38
rs145019158 Chr.17: 58351521-58351521 on Build GRCh38
rs771103557 Chr.17: 58351463-58351463 on Build GRCh38

View Full Product Details