Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54430890-54431157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTYH1
Entrez Gene ID
57348
Gene Name
tweety family member 1
Gene Aliases
-
UniGene
Hs.268728
Gene Chromosome Location
Chr.19: 54415431-54436719 on Build GRCh38
Transcripts
NM_001005367.2 NM_001201461.1 NM_020659.3 XM_017027025.1 XM_017027026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1590287 c.1075G>T p.G359* Chr.19: 54431141-54431141 on Build GRCh38
1590288 c.1030C>T p.Q344* Chr.19: 54430903-54430903 on Build GRCh38
5548232 c.1124-4delG p.? Chr.19: 54431095-54431095 on Build GRCh38
1683227 c.1062_1077del16 p.N355fs*>91 Chr.19: 54431128-54431143 on Build GRCh38
1001134 c.1030C>T p.Q344* Chr.19: 54430903-54430903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898323357 Chr.19: 54431029-54431029 on Build GRCh38
rs1050720369 Chr.19: 54431083-54431083 on Build GRCh38
rs529906657 Chr.19: 54430924-54430924 on Build GRCh38
rs1039659837 Chr.19: 54431027-54431028 on Build GRCh38
rs372871383 Chr.19: 54430922-54430922 on Build GRCh38
rs890857368 Chr.19: 54430949-54430950 on Build GRCh38
rs747886633 Chr.19: 54430929-54430930 on Build GRCh38
rs748892846 Chr.19: 54431120-54431120 on Build GRCh38
rs375960303 Chr.19: 54431029-54431029 on Build GRCh38
rs772111985 Chr.19: 54431062-54431062 on Build GRCh38
rs146138737 Chr.19: 54431122-54431122 on Build GRCh38
rs749060504 Chr.19: 54430943-54430943 on Build GRCh38
rs771760175 Chr.19: 54431049-54431058 on Build GRCh38
rs372640517 Chr.19: 54431027-54431027 on Build GRCh38
rs758937211 Chr.19: 54431102-54431102 on Build GRCh38
rs984852883 Chr.19: 54430995-54430995 on Build GRCh38
rs779884051 Chr.19: 54431119-54431119 on Build GRCh38
rs765759819 Chr.19: 54430919-54430919 on Build GRCh38
rs766991356 Chr.19: 54431109-54431109 on Build GRCh38
rs902311790 Chr.19: 54431108-54431108 on Build GRCh38
rs368694798 Chr.19: 54430978-54430978 on Build GRCh38
rs201932775 Chr.19: 54431069-54431069 on Build GRCh38
rs568548719 Chr.19: 54431029-54431030 on Build GRCh38
rs748053222 Chr.19: 54430957-54430957 on Build GRCh38
rs753145819 Chr.19: 54431099-54431099 on Build GRCh38
rs774169310 Chr.19: 54430953-54430953 on Build GRCh38
rs776797943 Chr.19: 54431084-54431084 on Build GRCh38
rs753195247 Chr.19: 54430920-54430920 on Build GRCh38
rs768522913 Chr.19: 54430952-54430952 on Build GRCh38
rs111737429 Chr.19: 54431014-54431014 on Build GRCh38
rs369258936 Chr.19: 54431079-54431079 on Build GRCh38
rs1046815060 Chr.19: 54431038-54431038 on Build GRCh38
rs201071440 Chr.19: 54431117-54431117 on Build GRCh38
rs976118257 Chr.19: 54431040-54431040 on Build GRCh38
rs189114469 Chr.19: 54430935-54430935 on Build GRCh38
rs923276699 Chr.19: 54431056-54431056 on Build GRCh38
rs755813502 Chr.19: 54430934-54430934 on Build GRCh38
rs765576262 Chr.19: 54431094-54431094 on Build GRCh38
rs977472106 Chr.19: 54430950-54430950 on Build GRCh38
rs755871014 Chr.19: 54431111-54431111 on Build GRCh38
rs914712410 Chr.19: 54430938-54430938 on Build GRCh38
rs113818416 Chr.19: 54431071-54431071 on Build GRCh38
rs951930796 Chr.19: 54430979-54430979 on Build GRCh38
rs958428766 Chr.19: 54431085-54431085 on Build GRCh38
rs933288005 Chr.19: 54430961-54430961 on Build GRCh38
rs1043448945 Chr.19: 54431107-54431107 on Build GRCh38
rs924701095 Chr.19: 54430960-54430960 on Build GRCh38
rs962029143 Chr.19: 54431028-54431028 on Build GRCh38
rs532280402 Chr.19: 54430987-54430987 on Build GRCh38
rs1002517306 Chr.19: 54431070-54431070 on Build GRCh38
rs896166743 Chr.19: 54431118-54431118 on Build GRCh38
rs759911976 Chr.19: 54431090-54431090 on Build GRCh38
rs942591729 Chr.19: 54431019-54431019 on Build GRCh38
rs999233551 Chr.19: 54431115-54431115 on Build GRCh38
rs755523718 Chr.19: 54430909-54430909 on Build GRCh38
rs552463201 Chr.19: 54430996-54430996 on Build GRCh38
rs959232248 Chr.19: 54430930-54430930 on Build GRCh38
rs1034898392 Chr.19: 54430933-54430933 on Build GRCh38

View Full Product Details