Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 3615750-3616011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKX
Entrez Gene ID
5613
Gene Name
protein kinase, X-linked
Gene Aliases
PKX1
UniGene
Hs.390788
Gene Chromosome Location
Chr.X: 3604343-3713634 on Build GRCh38
Transcripts
NM_005044.4

SNP Detail

SNP ID Chromosome Location
rs370229338 Chr.X: 3615850-3615850 on Build GRCh38
rs771312776 Chr.X: 3615855-3615855 on Build GRCh38
rs377672184 Chr.X: 3615779-3615779 on Build GRCh38
rs367895997 Chr.X: 3615818-3615818 on Build GRCh38
rs761705030 Chr.X: 3615916-3615916 on Build GRCh38
rs1031108093 Chr.X: 3615983-3615983 on Build GRCh38
rs138517585 Chr.X: 3615831-3615831 on Build GRCh38
rs771773396 Chr.X: 3615865-3615865 on Build GRCh38
rs900159954 Chr.X: 3615885-3615885 on Build GRCh38
rs765786646 Chr.X: 3615955-3615955 on Build GRCh38
rs191876092 Chr.X: 3615949-3615949 on Build GRCh38
rs41309657 Chr.X: 3615963-3615963 on Build GRCh38
rs753677394 Chr.X: 3615810-3615810 on Build GRCh38
rs2302751 Chr.X: 3615851-3615851 on Build GRCh38
rs868294807 Chr.X: 3615841-3615841 on Build GRCh38
rs752799338 Chr.X: 3615943-3615943 on Build GRCh38
rs147093558 Chr.X: 3615824-3615824 on Build GRCh38
rs141012473 Chr.X: 3615805-3615805 on Build GRCh38
rs752454112 Chr.X: 3615788-3615788 on Build GRCh38
rs778063938 Chr.X: 3615853-3615853 on Build GRCh38
rs765060540 Chr.X: 3615920-3615920 on Build GRCh38
rs201619179 Chr.X: 3615780-3615780 on Build GRCh38
rs753531997 Chr.X: 3615915-3615915 on Build GRCh38
rs751003297 Chr.X: 3615987-3615987 on Build GRCh38
rs760345858 Chr.X: 3615889-3615889 on Build GRCh38
rs778514933 Chr.X: 3615791-3615791 on Build GRCh38
rs992442369 Chr.X: 3615806-3615806 on Build GRCh38
rs2302750 Chr.X: 3615921-3615921 on Build GRCh38
rs374111523 Chr.X: 3615854-3615854 on Build GRCh38
rs751558573 Chr.X: 3615830-3615830 on Build GRCh38
rs763857663 Chr.X: 3615901-3615901 on Build GRCh38
rs781194944 Chr.X: 3615937-3615937 on Build GRCh38
rs141998823 Chr.X: 3615856-3615856 on Build GRCh38
rs41302609 Chr.X: 3615911-3615911 on Build GRCh38
rs869301143 Chr.X: 3615985-3615985 on Build GRCh38
rs372894839 Chr.X: 3615932-3615932 on Build GRCh38
rs150699468 Chr.X: 3615864-3615864 on Build GRCh38
rs775143744 Chr.X: 3615884-3615884 on Build GRCh38
rs771491214 Chr.X: 3615771-3615771 on Build GRCh38
rs756290607 Chr.X: 3615944-3615944 on Build GRCh38

View Full Product Details