Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 60571949-60572222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB2A
Entrez Gene ID
5862
Gene Name
RAB2A, member RAS oncogene family
Gene Aliases
LHX, RAB2
UniGene
Hs.369017
Gene Chromosome Location
Chr.8: 60516910-60623644 on Build GRCh38
Transcripts
NM_001242644.1 NM_002865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
606090 c.174G>T p.Q58H Chr.8: 60572101-60572101 on Build GRCh38
4529654 c.162G>C p.Q54H Chr.8: 60572089-60572089 on Build GRCh38
6226603 c.129C>T p.F43F Chr.8: 60572056-60572056 on Build GRCh38
6242205 c.172C>T p.Q58* Chr.8: 60572099-60572099 on Build GRCh38
369196 c.152A>T p.D51V Chr.8: 60572079-60572079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766778919 Chr.8: 60572004-60572004 on Build GRCh38
rs374493531 Chr.8: 60572028-60572028 on Build GRCh38
rs371695881 Chr.8: 60572144-60572144 on Build GRCh38
rs572990529 Chr.8: 60572018-60572018 on Build GRCh38
rs978462215 Chr.8: 60572130-60572130 on Build GRCh38
rs375665634 Chr.8: 60572022-60572022 on Build GRCh38
rs1023901635 Chr.8: 60572175-60572175 on Build GRCh38
rs186158413 Chr.8: 60572141-60572141 on Build GRCh38
rs752998204 Chr.8: 60572010-60572010 on Build GRCh38
rs1014145809 Chr.8: 60572173-60572173 on Build GRCh38
rs757985960 Chr.8: 60572029-60572029 on Build GRCh38
rs976954172 Chr.8: 60572057-60572057 on Build GRCh38
rs967525385 Chr.8: 60572113-60572113 on Build GRCh38
rs754408105 Chr.8: 60572131-60572131 on Build GRCh38
rs373858561 Chr.8: 60572009-60572009 on Build GRCh38
rs774263355 Chr.8: 60572127-60572127 on Build GRCh38
rs746795234 Chr.8: 60572038-60572038 on Build GRCh38
rs368402545 Chr.8: 60572056-60572056 on Build GRCh38
rs960869172 Chr.8: 60572170-60572170 on Build GRCh38
rs774656203 Chr.8: 60572002-60572002 on Build GRCh38
rs937604118 Chr.8: 60572193-60572193 on Build GRCh38
rs759653831 Chr.8: 60572008-60572008 on Build GRCh38
rs754755646 Chr.8: 60572040-60572040 on Build GRCh38
rs180926020 Chr.8: 60572027-60572027 on Build GRCh38
rs777141519 Chr.8: 60572155-60572155 on Build GRCh38
rs1020040727 Chr.8: 60572069-60572069 on Build GRCh38
rs771183016 Chr.8: 60572099-60572099 on Build GRCh38
rs779960307 Chr.8: 60572036-60572036 on Build GRCh38
rs561670645 Chr.8: 60572136-60572136 on Build GRCh38
rs761138248 Chr.8: 60571994-60571994 on Build GRCh38
rs991752373 Chr.8: 60572194-60572194 on Build GRCh38
rs1018120586 Chr.8: 60572047-60572047 on Build GRCh38
rs754273907 Chr.8: 60572024-60572024 on Build GRCh38
rs761227842 Chr.8: 60572013-60572013 on Build GRCh38
rs778561908 Chr.8: 60572159-60572159 on Build GRCh38
rs762281003 Chr.8: 60572165-60572165 on Build GRCh38
rs1035407166 Chr.8: 60572166-60572166 on Build GRCh38
rs781309549 Chr.8: 60572053-60572053 on Build GRCh38

View Full Product Details