Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158572625-158572898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RXFP1
Entrez Gene ID
59350
Gene Name
relaxin/insulin like family peptide receptor 1
Gene Aliases
LGR7, RXFPR1
UniGene
Hs.591686
Gene Chromosome Location
Chr.4: 158521711-158653372 on Build GRCh38
Transcripts
NM_001253727.1 NM_001253728.1 NM_001253729.1 NM_001253730.1 NM_001253732.1 NM_001253733.1 NM_021634.3 NR_045579.1 NR_045580.1 NR_045581.1 NR_045582.1 NR_045583.1 NR_045584.1 XM_011532174.1 XM_011532175.1 XM_011532176.1 XM_011532179.2 XM_017008517.1 XM_017008518.1 XM_017008519.1 XM_017008520.1 XM_017008521.1 XM_017008522.1 XM_017008523.1 XM_017008524.1 XM_017008525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4961369 c.187+1G>A p.? Chr.4: 158572836-158572836 on Build GRCh38
3781775 c.65G>A p.G22E Chr.4: 158572713-158572713 on Build GRCh38
6166682 c.97C>A p.P33T Chr.4: 158572745-158572745 on Build GRCh38
6770824 c.55C>A p.H19N Chr.4: 158572703-158572703 on Build GRCh38
6099558 c.174T>C p.D58D Chr.4: 158572822-158572822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770926999 Chr.4: 158572792-158572792 on Build GRCh38
rs529788116 Chr.4: 158572646-158572646 on Build GRCh38
rs748771167 Chr.4: 158572647-158572647 on Build GRCh38
rs1023336558 Chr.4: 158572783-158572783 on Build GRCh38
rs908497244 Chr.4: 158572753-158572753 on Build GRCh38
rs371883290 Chr.4: 158572682-158572682 on Build GRCh38
rs376578292 Chr.4: 158572777-158572777 on Build GRCh38
rs748690062 Chr.4: 158572661-158572661 on Build GRCh38
rs951851424 Chr.4: 158572793-158572793 on Build GRCh38
rs372906848 Chr.4: 158572819-158572819 on Build GRCh38
rs753574783 Chr.4: 158572802-158572802 on Build GRCh38
rs368783436 Chr.4: 158572795-158572795 on Build GRCh38
rs745880138 Chr.4: 158572681-158572682 on Build GRCh38
rs781226555 Chr.4: 158572861-158572861 on Build GRCh38
rs759489582 Chr.4: 158572798-158572798 on Build GRCh38
rs774207970 Chr.4: 158572797-158572797 on Build GRCh38
rs201512541 Chr.4: 158572651-158572651 on Build GRCh38
rs780532708 Chr.4: 158572737-158572737 on Build GRCh38
rs971079220 Chr.4: 158572705-158572705 on Build GRCh38
rs769807314 Chr.4: 158572866-158572866 on Build GRCh38
rs752478112 Chr.4: 158572805-158572805 on Build GRCh38
rs773339770 Chr.4: 158572877-158572877 on Build GRCh38
rs746265065 Chr.4: 158572715-158572715 on Build GRCh38
rs779248423 Chr.4: 158572685-158572685 on Build GRCh38
rs772369528 Chr.4: 158572731-158572731 on Build GRCh38
rs747315304 Chr.4: 158572859-158572859 on Build GRCh38
rs373380478 Chr.4: 158572719-158572720 on Build GRCh38
rs763670611 Chr.4: 158572809-158572809 on Build GRCh38
rs750932337 Chr.4: 158572838-158572838 on Build GRCh38
rs960491580 Chr.4: 158572730-158572730 on Build GRCh38
rs560540695 Chr.4: 158572645-158572645 on Build GRCh38
rs981346392 Chr.4: 158572747-158572747 on Build GRCh38
rs761727867 Chr.4: 158572790-158572790 on Build GRCh38
rs1038172550 Chr.4: 158572824-158572824 on Build GRCh38
rs200038291 Chr.4: 158572799-158572799 on Build GRCh38
rs748384362 Chr.4: 158572863-158572863 on Build GRCh38
rs559778296 Chr.4: 158572823-158572823 on Build GRCh38
rs758966867 Chr.4: 158572840-158572840 on Build GRCh38
rs747298846 Chr.4: 158572741-158572741 on Build GRCh38
rs990645240 Chr.4: 158572782-158572782 on Build GRCh38
rs867472717 Chr.4: 158572708-158572708 on Build GRCh38
rs768999539 Chr.4: 158572860-158572860 on Build GRCh38
rs140878326 Chr.4: 158572856-158572856 on Build GRCh38
rs915560356 Chr.4: 158572704-158572704 on Build GRCh38
rs760334929 Chr.4: 158572807-158572807 on Build GRCh38
rs768983121 Chr.4: 158572744-158572744 on Build GRCh38

View Full Product Details