Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130907872-130908145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX19
Entrez Gene ID
399979
Gene Name
sorting nexin 19
Gene Aliases
CHET8
UniGene
Hs.444024
Gene Chromosome Location
Chr.11: 130866254-130916488 on Build GRCh38
Transcripts
NM_001301089.1 NM_014758.2 XM_005271546.4 XM_011542819.2 XM_011542820.2 XM_011542821.2 XM_011542822.2 XM_011542824.2 XM_011542825.2 XM_011542826.2 XM_011542827.2 XM_017017725.1 XM_017017726.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1352642 c.2048C>T p.A683V Chr.11: 130908070-130908070 on Build GRCh38
203890 c.2072C>T p.A691V Chr.11: 130908046-130908046 on Build GRCh38
4417964 c.2127C>T p.T709T Chr.11: 130907991-130907991 on Build GRCh38
6269812 c.2089C>A p.P697T Chr.11: 130908029-130908029 on Build GRCh38
374974 c.2040G>T p.V680V Chr.11: 130908078-130908078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370854462 Chr.11: 130908105-130908105 on Build GRCh38
rs148900446 Chr.11: 130908000-130908000 on Build GRCh38
rs200362831 Chr.11: 130908011-130908011 on Build GRCh38
rs745969798 Chr.11: 130908103-130908103 on Build GRCh38
rs750331383 Chr.11: 130908056-130908056 on Build GRCh38
rs763114645 Chr.11: 130907916-130907916 on Build GRCh38
rs774982055 Chr.11: 130908010-130908010 on Build GRCh38
rs758291107 Chr.11: 130908061-130908061 on Build GRCh38
rs2276098 Chr.11: 130907991-130907991 on Build GRCh38
rs764778382 Chr.11: 130908045-130908045 on Build GRCh38
rs188916085 Chr.11: 130908117-130908117 on Build GRCh38
rs752370326 Chr.11: 130907944-130907944 on Build GRCh38
rs761117993 Chr.11: 130908025-130908025 on Build GRCh38
rs775953316 Chr.11: 130908017-130908017 on Build GRCh38
rs754803367 Chr.11: 130908072-130908072 on Build GRCh38
rs771109372 Chr.11: 130908080-130908080 on Build GRCh38
rs768049183 Chr.11: 130907938-130907938 on Build GRCh38
rs564430444 Chr.11: 130908003-130908003 on Build GRCh38
rs563224300 Chr.11: 130908037-130908037 on Build GRCh38
rs576544139 Chr.11: 130908048-130908048 on Build GRCh38
rs868221436 Chr.11: 130907954-130907954 on Build GRCh38
rs375635584 Chr.11: 130908034-130908034 on Build GRCh38
rs935528891 Chr.11: 130908027-130908027 on Build GRCh38
rs372569714 Chr.11: 130907997-130907997 on Build GRCh38
rs201117374 Chr.11: 130907958-130907958 on Build GRCh38
rs529043239 Chr.11: 130908077-130908077 on Build GRCh38
rs746075402 Chr.11: 130908092-130908092 on Build GRCh38
rs567953064 Chr.11: 130907956-130907956 on Build GRCh38
rs777849084 Chr.11: 130908075-130908075 on Build GRCh38
rs775000601 Chr.11: 130907932-130907932 on Build GRCh38
rs543634536 Chr.11: 130908012-130908012 on Build GRCh38
rs555298820 Chr.11: 130907907-130907907 on Build GRCh38
rs753233224 Chr.11: 130907972-130907972 on Build GRCh38
rs2276097 Chr.11: 130907918-130907918 on Build GRCh38
rs924194632 Chr.11: 130908013-130908013 on Build GRCh38
rs749256491 Chr.11: 130908007-130908007 on Build GRCh38
rs771106771 Chr.11: 130908008-130908008 on Build GRCh38
rs758168548 Chr.11: 130907996-130907996 on Build GRCh38
rs771923281 Chr.11: 130907902-130907902 on Build GRCh38
rs778820082 Chr.11: 130907985-130907985 on Build GRCh38
rs369718812 Chr.11: 130907960-130907960 on Build GRCh38
rs769886326 Chr.11: 130907906-130907906 on Build GRCh38
rs146517908 Chr.11: 130908068-130908068 on Build GRCh38
rs904778130 Chr.11: 130908028-130908028 on Build GRCh38
rs932115561 Chr.11: 130908119-130908119 on Build GRCh38
rs756797666 Chr.11: 130907982-130907982 on Build GRCh38
rs890157018 Chr.11: 130908078-130908078 on Build GRCh38
rs374087633 Chr.11: 130907904-130907904 on Build GRCh38
rs779099719 Chr.11: 130908094-130908094 on Build GRCh38
rs549908563 Chr.11: 130908023-130908023 on Build GRCh38
rs753594593 Chr.11: 130908035-130908035 on Build GRCh38
rs916271740 Chr.11: 130907979-130907979 on Build GRCh38
rs927749271 Chr.11: 130907909-130907909 on Build GRCh38

View Full Product Details