Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4533006-4533277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPNS2
Entrez Gene ID
124976
Gene Name
sphingolipid transporter 2
Gene Aliases
-
UniGene
Hs.22824
Gene Chromosome Location
Chr.17: 4498834-4539933 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4151642 c.1026C>T p.R342R Chr.17: 4533067-4533067 on Build GRCh38
4851195 c.1095C>G p.I365M Chr.17: 4533249-4533249 on Build GRCh38
247569 c.1068G>A p.P356P Chr.17: 4533109-4533109 on Build GRCh38
6998733 c.1118C>T p.T373M Chr.17: 4533272-4533272 on Build GRCh38
5706424 c.965C>T p.T322M Chr.17: 4533006-4533006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755120993 Chr.17: 4533110-4533110 on Build GRCh38
rs766194951 Chr.17: 4533135-4533136 on Build GRCh38
rs766105762 Chr.17: 4533173-4533173 on Build GRCh38
rs759354532 Chr.17: 4533225-4533225 on Build GRCh38
rs759665130 Chr.17: 4533042-4533042 on Build GRCh38
rs766373635 Chr.17: 4533109-4533109 on Build GRCh38
rs1053675335 Chr.17: 4533221-4533221 on Build GRCh38
rs780582867 Chr.17: 4533211-4533211 on Build GRCh38
rs868113905 Chr.17: 4533023-4533023 on Build GRCh38
rs746414162 Chr.17: 4533209-4533209 on Build GRCh38
rs369503350 Chr.17: 4533139-4533139 on Build GRCh38
rs779351260 Chr.17: 4533208-4533208 on Build GRCh38
rs775388707 Chr.17: 4533142-4533142 on Build GRCh38
rs780956983 Chr.17: 4533090-4533090 on Build GRCh38
rs765107938 Chr.17: 4533106-4533106 on Build GRCh38
rs375295357 Chr.17: 4533043-4533043 on Build GRCh38
rs754531478 Chr.17: 4533111-4533111 on Build GRCh38
rs764343600 Chr.17: 4533157-4533157 on Build GRCh38
rs773671931 Chr.17: 4533096-4533096 on Build GRCh38
rs760738107 Chr.17: 4533104-4533104 on Build GRCh38
rs752857169 Chr.17: 4533051-4533051 on Build GRCh38
rs752197156 Chr.17: 4533199-4533199 on Build GRCh38
rs372939841 Chr.17: 4533064-4533064 on Build GRCh38
rs559424699 Chr.17: 4533031-4533031 on Build GRCh38
rs963140825 Chr.17: 4533055-4533055 on Build GRCh38
rs767078306 Chr.17: 4533198-4533198 on Build GRCh38
rs201455640 Chr.17: 4533107-4533107 on Build GRCh38
rs755618446 Chr.17: 4533200-4533200 on Build GRCh38
rs200317055 Chr.17: 4533099-4533099 on Build GRCh38
rs61746307 Chr.17: 4533059-4533059 on Build GRCh38
rs758101824 Chr.17: 4533116-4533117 on Build GRCh38
rs773968730 Chr.17: 4533222-4533222 on Build GRCh38
rs377449141 Chr.17: 4533108-4533108 on Build GRCh38
rs368417189 Chr.17: 4533094-4533094 on Build GRCh38
rs1000428820 Chr.17: 4533088-4533088 on Build GRCh38
rs753526757 Chr.17: 4533206-4533206 on Build GRCh38
rs200092769 Chr.17: 4533027-4533027 on Build GRCh38
rs1036041932 Chr.17: 4533190-4533190 on Build GRCh38
rs201071887 Chr.17: 4533232-4533232 on Build GRCh38
rs767393853 Chr.17: 4533229-4533229 on Build GRCh38
rs750362032 Chr.17: 4533057-4533057 on Build GRCh38
rs371622226 Chr.17: 4533028-4533028 on Build GRCh38
rs780502709 Chr.17: 4533120-4533120 on Build GRCh38
rs767479513 Chr.17: 4533102-4533102 on Build GRCh38
rs771614086 Chr.17: 4533097-4533097 on Build GRCh38
rs746565744 Chr.17: 4533065-4533065 on Build GRCh38
rs947945778 Chr.17: 4533148-4533148 on Build GRCh38
rs762797449 Chr.17: 4533172-4533172 on Build GRCh38
rs747865152 Chr.17: 4533121-4533121 on Build GRCh38
rs201600154 Chr.17: 4533205-4533205 on Build GRCh38
rs747128774 Chr.17: 4533213-4533213 on Build GRCh38
rs182141708 Chr.17: 4533067-4533067 on Build GRCh38
rs199904911 Chr.17: 4533140-4533140 on Build GRCh38
rs1046644364 Chr.17: 4533168-4533168 on Build GRCh38
rs538437986 Chr.17: 4533220-4533220 on Build GRCh38
rs753103134 Chr.17: 4533195-4533201 on Build GRCh38
rs749710481 Chr.17: 4533133-4533133 on Build GRCh38
rs372798367 Chr.17: 4533134-4533134 on Build GRCh38
rs777545158 Chr.17: 4533122-4533122 on Build GRCh38
rs751641079 Chr.17: 4533038-4533038 on Build GRCh38
rs757131346 Chr.17: 4533052-4533052 on Build GRCh38
rs754832652 Chr.17: 4533197-4533197 on Build GRCh38
rs565424415 Chr.17: 4533068-4533068 on Build GRCh38
rs187158003 Chr.17: 4533216-4533216 on Build GRCh38
rs1053361978 Chr.17: 4533093-4533093 on Build GRCh38
rs768711756 Chr.17: 4533215-4533215 on Build GRCh38
rs757707153 Chr.17: 4533207-4533207 on Build GRCh38
rs373133643 Chr.17: 4533180-4533180 on Build GRCh38
rs770538003 Chr.17: 4533219-4533219 on Build GRCh38
rs763638535 Chr.17: 4533238-4533238 on Build GRCh38
rs775791130 Chr.17: 4533103-4533103 on Build GRCh38
rs904263442 Chr.17: 4533135-4533135 on Build GRCh38
rs529263790 Chr.17: 4533025-4533025 on Build GRCh38
rs370911335 Chr.17: 4533171-4533171 on Build GRCh38
rs772194236 Chr.17: 4533141-4533141 on Build GRCh38
rs546673107 Chr.17: 4533210-4533210 on Build GRCh38
rs759726200 Chr.17: 4533101-4533101 on Build GRCh38
rs575368818 Chr.17: 4533237-4533237 on Build GRCh38
rs760970111 Chr.17: 4533144-4533144 on Build GRCh38

View Full Product Details