Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63119139-63119384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A24
Entrez Gene ID
283238
Gene Name
solute carrier family 22 member 24
Gene Aliases
NET46
UniGene
Hs.375139
Gene Chromosome Location
Chr.11: 63079940-63144822 on Build GRCh38
Transcripts
NM_001136506.2 NM_173586.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4995018 c.592G>T p.V198F Chr.11: 63119250-63119250 on Build GRCh38
4995017 c.592G>T p.V198F Chr.11: 63119250-63119250 on Build GRCh38
4923490 c.618A>T p.A206A Chr.11: 63119224-63119224 on Build GRCh38
3998526 c.514C>G p.R172G Chr.11: 63119328-63119328 on Build GRCh38
4923489 c.618A>T p.A206A Chr.11: 63119224-63119224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147228662 Chr.11: 63119257-63119257 on Build GRCh38
rs763270233 Chr.11: 63119271-63119271 on Build GRCh38
rs777640983 Chr.11: 63119225-63119225 on Build GRCh38
rs777254687 Chr.11: 63119335-63119335 on Build GRCh38
rs750883393 Chr.11: 63119281-63119281 on Build GRCh38
rs764735365 Chr.11: 63119327-63119327 on Build GRCh38
rs776397966 Chr.11: 63119242-63119242 on Build GRCh38
rs747797825 Chr.11: 63119201-63119201 on Build GRCh38
rs748736693 Chr.11: 63119344-63119344 on Build GRCh38
rs913548653 Chr.11: 63119205-63119205 on Build GRCh38
rs768119348 Chr.11: 63119279-63119279 on Build GRCh38
rs370606508 Chr.11: 63119314-63119314 on Build GRCh38
rs557316228 Chr.11: 63119265-63119265 on Build GRCh38
rs778595562 Chr.11: 63119169-63119169 on Build GRCh38
rs557524759 Chr.11: 63119175-63119175 on Build GRCh38
rs1053567678 Chr.11: 63119183-63119183 on Build GRCh38
rs4963245 Chr.11: 63119328-63119328 on Build GRCh38
rs1045575534 Chr.11: 63119304-63119304 on Build GRCh38
rs74434787 Chr.11: 63119235-63119235 on Build GRCh38
rs116063135 Chr.11: 63119234-63119234 on Build GRCh38
rs746006880 Chr.11: 63119255-63119255 on Build GRCh38
rs373015794 Chr.11: 63119319-63119319 on Build GRCh38
rs867388193 Chr.11: 63119176-63119176 on Build GRCh38
rs781734003 Chr.11: 63119316-63119316 on Build GRCh38
rs746857513 Chr.11: 63119347-63119347 on Build GRCh38
rs911641800 Chr.11: 63119213-63119213 on Build GRCh38
rs888648064 Chr.11: 63119291-63119291 on Build GRCh38
rs117269586 Chr.11: 63119266-63119266 on Build GRCh38
rs945024633 Chr.11: 63119229-63119229 on Build GRCh38
rs193179908 Chr.11: 63119272-63119272 on Build GRCh38
rs550709193 Chr.11: 63119222-63119222 on Build GRCh38
rs906604172 Chr.11: 63119306-63119306 on Build GRCh38
rs556582100 Chr.11: 63119273-63119273 on Build GRCh38

View Full Product Details