Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9896322-9896567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHROOM2
Entrez Gene ID
357
Gene Name
shroom family member 2
Gene Aliases
APXL, HSAPXL
UniGene
Hs.567236
Gene Chromosome Location
Chr.X: 9786406-9949443 on Build GRCh38
Transcripts
NM_001649.3 XM_005274500.4

SNP Detail

SNP ID Chromosome Location
rs910165925 Chr.X: 9896462-9896462 on Build GRCh38
rs376124648 Chr.X: 9896466-9896466 on Build GRCh38
rs1051476639 Chr.X: 9896389-9896389 on Build GRCh38
rs778519994 Chr.X: 9896409-9896409 on Build GRCh38
rs746770133 Chr.X: 9896477-9896477 on Build GRCh38
rs758331579 Chr.X: 9896459-9896459 on Build GRCh38
rs745671156 Chr.X: 9896421-9896421 on Build GRCh38
rs753343617 Chr.X: 9896480-9896480 on Build GRCh38
rs148686929 Chr.X: 9896511-9896511 on Build GRCh38
rs933084306 Chr.X: 9896512-9896512 on Build GRCh38
rs371590825 Chr.X: 9896476-9896476 on Build GRCh38
rs1035679943 Chr.X: 9896450-9896450 on Build GRCh38
rs141630186 Chr.X: 9896408-9896408 on Build GRCh38
rs762947966 Chr.X: 9896538-9896538 on Build GRCh38
rs934975666 Chr.X: 9896535-9896535 on Build GRCh38
rs368355034 Chr.X: 9896519-9896519 on Build GRCh38
rs773234401 Chr.X: 9896439-9896439 on Build GRCh38
rs61744804 Chr.X: 9896361-9896361 on Build GRCh38
rs984500046 Chr.X: 9896449-9896449 on Build GRCh38
rs1007611611 Chr.X: 9896398-9896398 on Build GRCh38
rs73474584 Chr.X: 9896481-9896481 on Build GRCh38
rs144110998 Chr.X: 9896482-9896482 on Build GRCh38
rs987200110 Chr.X: 9896518-9896518 on Build GRCh38
rs141610350 Chr.X: 9896369-9896369 on Build GRCh38
rs767943035 Chr.X: 9896467-9896467 on Build GRCh38
rs778477756 Chr.X: 9896395-9896395 on Build GRCh38
rs150083189 Chr.X: 9896401-9896401 on Build GRCh38
rs950708993 Chr.X: 9896456-9896456 on Build GRCh38
rs202190752 Chr.X: 9896509-9896509 on Build GRCh38
rs774052430 Chr.X: 9896347-9896347 on Build GRCh38
rs374462030 Chr.X: 9896510-9896510 on Build GRCh38
rs61739700 Chr.X: 9896366-9896366 on Build GRCh38
rs12006769 Chr.X: 9896403-9896403 on Build GRCh38
rs766620817 Chr.X: 9896394-9896394 on Build GRCh38
rs570983388 Chr.X: 9896373-9896373 on Build GRCh38
rs373190071 Chr.X: 9896425-9896425 on Build GRCh38
rs776574068 Chr.X: 9896522-9896522 on Build GRCh38
rs200542562 Chr.X: 9896404-9896404 on Build GRCh38
rs982172106 Chr.X: 9896438-9896438 on Build GRCh38
rs755337260 Chr.X: 9896397-9896397 on Build GRCh38
rs146225710 Chr.X: 9896443-9896443 on Build GRCh38
rs760581631 Chr.X: 9896436-9896436 on Build GRCh38
rs775430947 Chr.X: 9896429-9896429 on Build GRCh38
rs750497119 Chr.X: 9896374-9896374 on Build GRCh38
rs763126396 Chr.X: 9896387-9896387 on Build GRCh38
rs768608149 Chr.X: 9896437-9896437 on Build GRCh38
rs777570154 Chr.X: 9896470-9896470 on Build GRCh38
rs137979992 Chr.X: 9896446-9896446 on Build GRCh38
rs745478150 Chr.X: 9896496-9896496 on Build GRCh38

View Full Product Details