Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 105278315-105278546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PREP
Entrez Gene ID
5550
Gene Name
prolyl endopeptidase
Gene Aliases
PE, PEP
UniGene
Hs.436564
Gene Chromosome Location
Chr.6: 105277567-105403124 on Build GRCh38
Transcripts
NM_002726.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3154160 c.1892C>A p.P631Q Chr.6: 105278385-105278385 on Build GRCh38
1071720 c.1929C>T p.R643R Chr.6: 105278348-105278348 on Build GRCh38
4886904 c.1895C>A p.S632Y Chr.6: 105278382-105278382 on Build GRCh38
7107920 c.1870G>A p.E624K Chr.6: 105278407-105278407 on Build GRCh38
3154159 c.1893G>A p.P631P Chr.6: 105278384-105278384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764693264 Chr.6: 105278366-105278366 on Build GRCh38
rs775679233 Chr.6: 105278481-105278481 on Build GRCh38
rs745753367 Chr.6: 105278468-105278469 on Build GRCh38
rs779100440 Chr.6: 105278478-105278478 on Build GRCh38
rs781208545 Chr.6: 105278455-105278455 on Build GRCh38
rs750170373 Chr.6: 105278419-105278419 on Build GRCh38
rs768988387 Chr.6: 105278490-105278490 on Build GRCh38
rs772490679 Chr.6: 105278350-105278350 on Build GRCh38
rs1037840936 Chr.6: 105278502-105278502 on Build GRCh38
rs186512128 Chr.6: 105278447-105278447 on Build GRCh38
rs764920544 Chr.6: 105278384-105278384 on Build GRCh38
rs150938968 Chr.6: 105278338-105278338 on Build GRCh38
rs757334021 Chr.6: 105278459-105278459 on Build GRCh38
rs377412305 Chr.6: 105278351-105278351 on Build GRCh38
rs898855293 Chr.6: 105278341-105278341 on Build GRCh38
rs761128853 Chr.6: 105278354-105278354 on Build GRCh38
rs747422563 Chr.6: 105278484-105278484 on Build GRCh38
rs922965275 Chr.6: 105278428-105278428 on Build GRCh38
rs1009037501 Chr.6: 105278394-105278394 on Build GRCh38
rs746144989 Chr.6: 105278480-105278480 on Build GRCh38
rs942191599 Chr.6: 105278497-105278497 on Build GRCh38
rs749307832 Chr.6: 105278457-105278457 on Build GRCh38
rs746163142 Chr.6: 105278349-105278349 on Build GRCh38
rs372879716 Chr.6: 105278339-105278339 on Build GRCh38
rs754826892 Chr.6: 105278449-105278449 on Build GRCh38
rs769652411 Chr.6: 105278481-105278482 on Build GRCh38
rs370122132 Chr.6: 105278426-105278426 on Build GRCh38
rs149696511 Chr.6: 105278367-105278367 on Build GRCh38
rs761426091 Chr.6: 105278369-105278369 on Build GRCh38
rs766133270 Chr.6: 105278438-105278438 on Build GRCh38
rs149186054 Chr.6: 105278348-105278348 on Build GRCh38

View Full Product Details