Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 124031809-124032082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM229A
Entrez Gene ID
730130
Gene Name
transmembrane protein 229A
Gene Aliases
-
UniGene
Hs.450249
Gene Chromosome Location
Chr.7: 124030916-124033469 on Build GRCh38
Transcripts
NM_001136002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6515533 c.1108G>T p.V370L Chr.7: 124031896-124031896 on Build GRCh38
6515535 c.1047C>T p.I349I Chr.7: 124031957-124031957 on Build GRCh38
4921213 c.990G>A p.T330T Chr.7: 124032014-124032014 on Build GRCh38
5910597 c.1033T>G p.F345V Chr.7: 124031971-124031971 on Build GRCh38
1447738 c.993C>T p.C331C Chr.7: 124032011-124032011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008455249 Chr.7: 124032019-124032019 on Build GRCh38
rs755150268 Chr.7: 124031908-124031908 on Build GRCh38
rs746203949 Chr.7: 124031838-124031839 on Build GRCh38
rs1045323507 Chr.7: 124031893-124031893 on Build GRCh38
rs1027660035 Chr.7: 124032043-124032043 on Build GRCh38
rs749775845 Chr.7: 124031884-124031884 on Build GRCh38
rs769312335 Chr.7: 124031897-124031897 on Build GRCh38
rs555292779 Chr.7: 124031844-124031844 on Build GRCh38
rs995721186 Chr.7: 124032007-124032007 on Build GRCh38
rs754024714 Chr.7: 124031894-124031894 on Build GRCh38
rs200117879 Chr.7: 124032053-124032053 on Build GRCh38
rs182026849 Chr.7: 124031863-124031863 on Build GRCh38
rs939536641 Chr.7: 124031910-124031910 on Build GRCh38
rs1007559773 Chr.7: 124032050-124032050 on Build GRCh38
rs1056622890 Chr.7: 124031926-124031926 on Build GRCh38
rs748467260 Chr.7: 124031970-124031970 on Build GRCh38
rs765537937 Chr.7: 124032035-124032035 on Build GRCh38
rs563805340 Chr.7: 124032049-124032049 on Build GRCh38
rs887746238 Chr.7: 124032046-124032046 on Build GRCh38
rs896501385 Chr.7: 124031850-124031850 on Build GRCh38
rs1017573776 Chr.7: 124032056-124032056 on Build GRCh38
rs755796134 Chr.7: 124031850-124031851 on Build GRCh38
rs756122599 Chr.7: 124031858-124031858 on Build GRCh38
rs780324135 Chr.7: 124031879-124031879 on Build GRCh38
rs781261496 Chr.7: 124031836-124031836 on Build GRCh38
rs990239379 Chr.7: 124031842-124031842 on Build GRCh38
rs562410649 Chr.7: 124031839-124031842 on Build GRCh38
rs954345192 Chr.7: 124032017-124032017 on Build GRCh38
rs540636871 Chr.7: 124032010-124032010 on Build GRCh38
rs898570494 Chr.7: 124031999-124031999 on Build GRCh38
rs1001764585 Chr.7: 124031883-124031883 on Build GRCh38

View Full Product Details