Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151066472-151066729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC4A2
Entrez Gene ID
6522
Gene Name
solute carrier family 4 member 2
Gene Aliases
AE2, BND3L, EPB3L1, HKB3, NBND3
UniGene
Hs.647069
Gene Chromosome Location
Chr.7: 151058212-151076527 on Build GRCh38
Transcripts
NM_001199692.1 NM_001199693.1 NM_001199694.1 NM_003040.3 XM_006716094.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6189339 c.579-1G>C p.? Chr.7: 151066516-151066516 on Build GRCh38
1622563 c.580A>G p.T194A Chr.7: 151066518-151066518 on Build GRCh38
5723908 c.716G>A p.R239K Chr.7: 151066654-151066654 on Build GRCh38
7426407 c.701A>T p.N234I Chr.7: 151066639-151066639 on Build GRCh38
224950 c.767C>T p.P256L Chr.7: 151066705-151066705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199537782 Chr.7: 151066534-151066534 on Build GRCh38
rs1011219539 Chr.7: 151066626-151066626 on Build GRCh38
rs150330052 Chr.7: 151066529-151066534 on Build GRCh38
rs35360355 Chr.7: 151066497-151066497 on Build GRCh38
rs754802714 Chr.7: 151066563-151066563 on Build GRCh38
rs201747636 Chr.7: 151066576-151066576 on Build GRCh38
rs775944006 Chr.7: 151066577-151066577 on Build GRCh38
rs758046342 Chr.7: 151066544-151066549 on Build GRCh38
rs752478649 Chr.7: 151066698-151066698 on Build GRCh38
rs754930320 Chr.7: 151066683-151066683 on Build GRCh38
rs942786651 Chr.7: 151066673-151066673 on Build GRCh38
rs957017539 Chr.7: 151066620-151066620 on Build GRCh38
rs755030840 Chr.7: 151066703-151066703 on Build GRCh38
rs747991446 Chr.7: 151066565-151066565 on Build GRCh38
rs750110371 Chr.7: 151066595-151066595 on Build GRCh38
rs2229549 Chr.7: 151066568-151066568 on Build GRCh38
rs762099826 Chr.7: 151066581-151066581 on Build GRCh38
rs982341791 Chr.7: 151066592-151066592 on Build GRCh38
rs770274600 Chr.7: 151066498-151066498 on Build GRCh38
rs375977483 Chr.7: 151066584-151066584 on Build GRCh38
rs542333563 Chr.7: 151066572-151066572 on Build GRCh38
rs35506863 Chr.7: 151066553-151066558 on Build GRCh38
rs778805240 Chr.7: 151066564-151066564 on Build GRCh38
rs763497586 Chr.7: 151066578-151066578 on Build GRCh38
rs755639002 Chr.7: 151066552-151066552 on Build GRCh38
rs557013894 Chr.7: 151066535-151066535 on Build GRCh38
rs755066078 Chr.7: 151066534-151066535 on Build GRCh38
rs2229548 Chr.7: 151066553-151066553 on Build GRCh38
rs2229551 Chr.7: 151066543-151066543 on Build GRCh38
rs766040512 Chr.7: 151066661-151066661 on Build GRCh38
rs571570942 Chr.7: 151066514-151066514 on Build GRCh38
rs997466181 Chr.7: 151066519-151066519 on Build GRCh38
rs369447066 Chr.7: 151066709-151066709 on Build GRCh38
rs753365288 Chr.7: 151066575-151066575 on Build GRCh38
rs999056267 Chr.7: 151066594-151066594 on Build GRCh38
rs374531279 Chr.7: 151066543-151066548 on Build GRCh38
rs760204657 Chr.7: 151066599-151066599 on Build GRCh38
rs761405189 Chr.7: 151066546-151066546 on Build GRCh38
rs970939987 Chr.7: 151066588-151066588 on Build GRCh38
rs537018427 Chr.7: 151066524-151066524 on Build GRCh38
rs369100379 Chr.7: 151066549-151066549 on Build GRCh38
rs774633051 Chr.7: 151066580-151066580 on Build GRCh38
rs1031328236 Chr.7: 151066600-151066600 on Build GRCh38
rs375839693 Chr.7: 151066547-151066547 on Build GRCh38
rs778995423 Chr.7: 151066696-151066696 on Build GRCh38
rs17852578 Chr.7: 151066603-151066603 on Build GRCh38
rs753426372 Chr.7: 151066674-151066674 on Build GRCh38
rs781279800 Chr.7: 151066570-151066570 on Build GRCh38
rs745870637 Chr.7: 151066528-151066529 on Build GRCh38
rs984341382 Chr.7: 151066622-151066622 on Build GRCh38
rs902364864 Chr.7: 151066541-151066541 on Build GRCh38

View Full Product Details