Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20891617-20891856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASE3
Entrez Gene ID
6037
Gene Name
ribonuclease A family member 3
Gene Aliases
ECP, RAF1, RNS3
UniGene
Hs.73839
Gene Chromosome Location
Chr.14: 20891403-20892348 on Build GRCh38
Transcripts
NM_002935.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6767075 c.86C>T p.P29L Chr.14: 20891772-20891772 on Build GRCh38
432817 c.12A>G p.K4K Chr.14: 20891698-20891698 on Build GRCh38
6521309 c.140C>A p.P47H Chr.14: 20891826-20891826 on Build GRCh38
6767077 c.143C>A p.P48H Chr.14: 20891829-20891829 on Build GRCh38
6140025 c.145C>G p.R49G Chr.14: 20891831-20891831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142084658 Chr.14: 20891824-20891824 on Build GRCh38
rs2233859 Chr.14: 20891649-20891649 on Build GRCh38
rs756609160 Chr.14: 20891718-20891718 on Build GRCh38
rs752773252 Chr.14: 20891716-20891716 on Build GRCh38
rs755183918 Chr.14: 20891699-20891699 on Build GRCh38
rs749108905 Chr.14: 20891782-20891783 on Build GRCh38
rs768163273 Chr.14: 20891653-20891653 on Build GRCh38
rs191323095 Chr.14: 20891667-20891667 on Build GRCh38
rs760867669 Chr.14: 20891813-20891813 on Build GRCh38
rs762792315 Chr.14: 20891771-20891771 on Build GRCh38
rs200170777 Chr.14: 20891829-20891829 on Build GRCh38
rs770621900 Chr.14: 20891798-20891798 on Build GRCh38
rs778394298 Chr.14: 20891731-20891731 on Build GRCh38
rs201435792 Chr.14: 20891807-20891807 on Build GRCh38
rs371809756 Chr.14: 20891784-20891784 on Build GRCh38
rs747737990 Chr.14: 20891750-20891750 on Build GRCh38
rs374635539 Chr.14: 20891741-20891741 on Build GRCh38
rs558397403 Chr.14: 20891645-20891645 on Build GRCh38
rs202232277 Chr.14: 20891828-20891828 on Build GRCh38
rs148601585 Chr.14: 20891774-20891774 on Build GRCh38
rs147642716 Chr.14: 20891659-20891659 on Build GRCh38
rs144042562 Chr.14: 20891814-20891814 on Build GRCh38
rs776152400 Chr.14: 20891747-20891747 on Build GRCh38
rs749747826 Chr.14: 20891738-20891738 on Build GRCh38
rs561660328 Chr.14: 20891821-20891821 on Build GRCh38
rs759859690 Chr.14: 20891785-20891785 on Build GRCh38
rs766069912 Chr.14: 20891689-20891689 on Build GRCh38
rs1029249479 Chr.14: 20891659-20891662 on Build GRCh38
rs149500970 Chr.14: 20891713-20891713 on Build GRCh38
rs775879390 Chr.14: 20891745-20891745 on Build GRCh38
rs762853405 Chr.14: 20891677-20891677 on Build GRCh38
rs765183900 Chr.14: 20891665-20891665 on Build GRCh38
rs867802498 Chr.14: 20891735-20891735 on Build GRCh38
rs751894020 Chr.14: 20891690-20891690 on Build GRCh38
rs762144004 Chr.14: 20891731-20891732 on Build GRCh38
rs140556531 Chr.14: 20891835-20891835 on Build GRCh38
rs749325429 Chr.14: 20891816-20891816 on Build GRCh38
rs767807631 Chr.14: 20891806-20891806 on Build GRCh38
rs368925772 Chr.14: 20891752-20891752 on Build GRCh38
rs188802084 Chr.14: 20891657-20891657 on Build GRCh38
rs774318039 Chr.14: 20891838-20891838 on Build GRCh38
rs371854422 Chr.14: 20891764-20891764 on Build GRCh38
rs779182743 Chr.14: 20891742-20891742 on Build GRCh38

View Full Product Details