Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142314129-142314402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRY4
Entrez Gene ID
81848
Gene Name
sprouty RTK signaling antagonist 4
Gene Aliases
HH17
UniGene
Hs.323308
Gene Chromosome Location
Chr.5: 142310427-142325055 on Build GRCh38
Transcripts
NM_001127496.1 NM_001293289.1 NM_001293290.1 NM_030964.3 XM_011537685.2 XM_017009910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5565442 c.879C>T p.R293R Chr.5: 142314299-142314299 on Build GRCh38
6102487 c.923C>G p.A308G Chr.5: 142314255-142314255 on Build GRCh38
6789359 c.856C>T p.R286C Chr.5: 142314322-142314322 on Build GRCh38
4631000 c.855G>T p.Q285H Chr.5: 142314323-142314323 on Build GRCh38
6102486 c.795C>T p.S265S Chr.5: 142314383-142314383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144313996 Chr.5: 142314163-142314163 on Build GRCh38
rs767152468 Chr.5: 142314231-142314231 on Build GRCh38
rs745342448 Chr.5: 142314212-142314213 on Build GRCh38
rs553491984 Chr.5: 142314276-142314276 on Build GRCh38
rs772065888 Chr.5: 142314181-142314181 on Build GRCh38
rs530411114 Chr.5: 142314275-142314275 on Build GRCh38
rs568658303 Chr.5: 142314289-142314289 on Build GRCh38
rs901907169 Chr.5: 142314326-142314326 on Build GRCh38
rs200364529 Chr.5: 142314337-142314337 on Build GRCh38
rs781714403 Chr.5: 142314362-142314362 on Build GRCh38
rs758080279 Chr.5: 142314283-142314283 on Build GRCh38
rs1046862683 Chr.5: 142314354-142314354 on Build GRCh38
rs372597519 Chr.5: 142314363-142314363 on Build GRCh38
rs766027202 Chr.5: 142314353-142314353 on Build GRCh38
rs753341165 Chr.5: 142314187-142314187 on Build GRCh38
rs148661341 Chr.5: 142314247-142314247 on Build GRCh38
rs763268380 Chr.5: 142314226-142314226 on Build GRCh38
rs747414690 Chr.5: 142314203-142314203 on Build GRCh38
rs768461830 Chr.5: 142314321-142314321 on Build GRCh38
rs756704150 Chr.5: 142314370-142314370 on Build GRCh38
rs375436542 Chr.5: 142314368-142314368 on Build GRCh38
rs764666555 Chr.5: 142314338-142314338 on Build GRCh38
rs145360326 Chr.5: 142314383-142314383 on Build GRCh38
rs376107467 Chr.5: 142314197-142314197 on Build GRCh38
rs748212197 Chr.5: 142314207-142314207 on Build GRCh38
rs200989792 Chr.5: 142314303-142314303 on Build GRCh38
rs377690087 Chr.5: 142314249-142314249 on Build GRCh38
rs749414864 Chr.5: 142314308-142314308 on Build GRCh38
rs778203577 Chr.5: 142314378-142314378 on Build GRCh38
rs778515546 Chr.5: 142314191-142314191 on Build GRCh38
rs769196353 Chr.5: 142314205-142314205 on Build GRCh38
rs758646143 Chr.5: 142314198-142314198 on Build GRCh38
rs749355008 Chr.5: 142314379-142314379 on Build GRCh38
rs756799416 Chr.5: 142314190-142314190 on Build GRCh38
rs377361633 Chr.5: 142314159-142314159 on Build GRCh38
rs139736145 Chr.5: 142314314-142314314 on Build GRCh38
rs770052498 Chr.5: 142314212-142314212 on Build GRCh38
rs775419201 Chr.5: 142314224-142314224 on Build GRCh38
rs531177415 Chr.5: 142314295-142314295 on Build GRCh38
rs777347198 Chr.5: 142314206-142314206 on Build GRCh38
rs887055648 Chr.5: 142314365-142314365 on Build GRCh38
rs751223130 Chr.5: 142314355-142314355 on Build GRCh38
rs147431622 Chr.5: 142314269-142314269 on Build GRCh38
rs748690914 Chr.5: 142314300-142314300 on Build GRCh38
rs781759921 Chr.5: 142314298-142314298 on Build GRCh38
rs779627371 Chr.5: 142314288-142314288 on Build GRCh38
rs906970597 Chr.5: 142314271-142314271 on Build GRCh38
rs762439277 Chr.5: 142314344-142314344 on Build GRCh38
rs775370197 Chr.5: 142314244-142314244 on Build GRCh38
rs951277676 Chr.5: 142314177-142314177 on Build GRCh38
rs201902270 Chr.5: 142314264-142314264 on Build GRCh38
rs749933707 Chr.5: 142314343-142314343 on Build GRCh38
rs367842050 Chr.5: 142314309-142314309 on Build GRCh38
rs201009569 Chr.5: 142314382-142314382 on Build GRCh38
rs755349527 Chr.5: 142314357-142314357 on Build GRCh38
rs773430976 Chr.5: 142314304-142314304 on Build GRCh38
rs528463342 Chr.5: 142314202-142314202 on Build GRCh38
rs771409068 Chr.5: 142314223-142314223 on Build GRCh38
rs569771989 Chr.5: 142314322-142314322 on Build GRCh38
rs75645175 Chr.5: 142314166-142314166 on Build GRCh38
rs756083952 Chr.5: 142314172-142314172 on Build GRCh38
rs774476082 Chr.5: 142314310-142314310 on Build GRCh38
rs142439525 Chr.5: 142314268-142314268 on Build GRCh38

View Full Product Details