Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12158055-12158280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC61A2
Entrez Gene ID
55176
Gene Name
Sec61 translocon alpha 2 subunit
Gene Aliases
-
UniGene
Hs.112955
Gene Chromosome Location
Chr.10: 12129641-12169958 on Build GRCh38
Transcripts
NM_001142627.2 NM_001142628.1 NM_018144.3 NR_024576.2 NR_024577.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5483439 c.931C>T p.R311* Chr.10: 12158061-12158061 on Build GRCh38
1128122 c.975+1G>A p.? Chr.10: 12158106-12158106 on Build GRCh38
6460932 c.962T>C p.L321P Chr.10: 12158092-12158092 on Build GRCh38
3723498 c.949T>C p.L317L Chr.10: 12158079-12158079 on Build GRCh38
4724606 c.932G>A p.R311Q Chr.10: 12158062-12158062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748194883 Chr.10: 12158115-12158115 on Build GRCh38
rs544615309 Chr.10: 12158186-12158186 on Build GRCh38
rs1017795348 Chr.10: 12158223-12158223 on Build GRCh38
rs981906800 Chr.10: 12158090-12158090 on Build GRCh38
rs914868512 Chr.10: 12158137-12158137 on Build GRCh38
rs146827112 Chr.10: 12158088-12158088 on Build GRCh38
rs4750175 Chr.10: 12158126-12158126 on Build GRCh38
rs377551011 Chr.10: 12158150-12158150 on Build GRCh38
rs1049877411 Chr.10: 12158190-12158190 on Build GRCh38
rs773503453 Chr.10: 12158127-12158127 on Build GRCh38
rs964132645 Chr.10: 12158226-12158226 on Build GRCh38
rs564781824 Chr.10: 12158219-12158219 on Build GRCh38
rs766211718 Chr.10: 12158134-12158134 on Build GRCh38
rs1043006522 Chr.10: 12158158-12158158 on Build GRCh38
rs752308866 Chr.10: 12158157-12158157 on Build GRCh38
rs770123874 Chr.10: 12158106-12158109 on Build GRCh38
rs760744118 Chr.10: 12158128-12158128 on Build GRCh38
rs1007719890 Chr.10: 12158194-12158194 on Build GRCh38
rs759613997 Chr.10: 12158149-12158149 on Build GRCh38
rs952164935 Chr.10: 12158179-12158179 on Build GRCh38
rs998743454 Chr.10: 12158185-12158185 on Build GRCh38
rs61731911 Chr.10: 12158093-12158093 on Build GRCh38
rs776278392 Chr.10: 12158136-12158136 on Build GRCh38
rs897836252 Chr.10: 12158105-12158105 on Build GRCh38

View Full Product Details