Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57497678-57497949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC43A1
Entrez Gene ID
8501
Gene Name
solute carrier family 43 member 1
Gene Aliases
LAT3, PB39, POV1, R00504
UniGene
Hs.591952
Gene Chromosome Location
Chr.11: 57484531-57515954 on Build GRCh38
Transcripts
NM_001198810.1 NM_003627.5 XM_005274358.4 XM_011545320.2 XM_011545321.2 XM_011545322.1 XM_017018451.1 XM_017018452.1 XM_017018453.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5678079 c.558G>T p.K186N Chr.11: 57497753-57497753 on Build GRCh38
5422356 c.558+4T>C p.? Chr.11: 57497749-57497749 on Build GRCh38
5665653 c.539T>C p.I180T Chr.11: 57497772-57497772 on Build GRCh38
7323619 c.495C>G p.S165S Chr.11: 57497816-57497816 on Build GRCh38
4567046 c.546_547CC>TT p.P183S Chr.11: 57497764-57497765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758157032 Chr.11: 57497726-57497726 on Build GRCh38
rs779788161 Chr.11: 57497736-57497736 on Build GRCh38
rs772137909 Chr.11: 57497829-57497829 on Build GRCh38
rs573863329 Chr.11: 57497718-57497718 on Build GRCh38
rs775618013 Chr.11: 57497834-57497834 on Build GRCh38
rs200312455 Chr.11: 57497800-57497800 on Build GRCh38
rs771377865 Chr.11: 57497802-57497802 on Build GRCh38
rs114467256 Chr.11: 57497768-57497768 on Build GRCh38
rs779323542 Chr.11: 57497820-57497820 on Build GRCh38
rs775770599 Chr.11: 57497764-57497764 on Build GRCh38
rs372034232 Chr.11: 57497858-57497858 on Build GRCh38
rs1034902501 Chr.11: 57497872-57497872 on Build GRCh38
rs138217743 Chr.11: 57497697-57497697 on Build GRCh38
rs181597007 Chr.11: 57497716-57497716 on Build GRCh38
rs376203144 Chr.11: 57497719-57497719 on Build GRCh38
rs936288743 Chr.11: 57497883-57497883 on Build GRCh38
rs2581922 Chr.11: 57497927-57497927 on Build GRCh38
rs977471183 Chr.11: 57497840-57497840 on Build GRCh38
rs201615562 Chr.11: 57497740-57497740 on Build GRCh38
rs756761958 Chr.11: 57497709-57497709 on Build GRCh38
rs753813477 Chr.11: 57497707-57497707 on Build GRCh38
rs951972809 Chr.11: 57497750-57497750 on Build GRCh38
rs746007553 Chr.11: 57497821-57497821 on Build GRCh38
rs374222722 Chr.11: 57497882-57497882 on Build GRCh38
rs368999945 Chr.11: 57497884-57497884 on Build GRCh38
rs562167576 Chr.11: 57497749-57497749 on Build GRCh38
rs749771090 Chr.11: 57497745-57497745 on Build GRCh38
rs749562185 Chr.11: 57497814-57497814 on Build GRCh38
rs760457060 Chr.11: 57497766-57497766 on Build GRCh38
rs767034024 Chr.11: 57497885-57497885 on Build GRCh38
rs760415134 Chr.11: 57497905-57497905 on Build GRCh38
rs145035847 Chr.11: 57497813-57497813 on Build GRCh38
rs897542401 Chr.11: 57497710-57497710 on Build GRCh38
rs1033552508 Chr.11: 57497769-57497769 on Build GRCh38
rs759939897 Chr.11: 57497863-57497864 on Build GRCh38
rs542813456 Chr.11: 57497806-57497806 on Build GRCh38
rs757557551 Chr.11: 57497889-57497889 on Build GRCh38
rs141426221 Chr.11: 57497786-57497786 on Build GRCh38
rs753772642 Chr.11: 57497782-57497782 on Build GRCh38
rs768712508 Chr.11: 57497845-57497845 on Build GRCh38
rs776520986 Chr.11: 57497851-57497851 on Build GRCh38
rs757925989 Chr.11: 57497790-57497790 on Build GRCh38
rs777149073 Chr.11: 57497728-57497731 on Build GRCh38
rs753108204 Chr.11: 57497877-57497878 on Build GRCh38
rs756474417 Chr.11: 57497809-57497809 on Build GRCh38
rs375259903 Chr.11: 57497864-57497864 on Build GRCh38
rs770180709 Chr.11: 57497742-57497742 on Build GRCh38
rs754231446 Chr.11: 57497886-57497886 on Build GRCh38
rs762617034 Chr.11: 57497865-57497865 on Build GRCh38
rs760933269 Chr.11: 57497842-57497842 on Build GRCh38
rs573386965 Chr.11: 57497698-57497698 on Build GRCh38
rs765881193 Chr.11: 57497881-57497881 on Build GRCh38
rs761636094 Chr.11: 57497857-57497857 on Build GRCh38
rs759570173 Chr.11: 57497756-57497756 on Build GRCh38
rs761833502 Chr.11: 57497784-57497784 on Build GRCh38
rs760463967 Chr.11: 57497703-57497703 on Build GRCh38
rs550877436 Chr.11: 57497819-57497819 on Build GRCh38
rs763999185 Chr.11: 57497704-57497704 on Build GRCh38
rs778209582 Chr.11: 57497744-57497744 on Build GRCh38
rs147862003 Chr.11: 57497785-57497785 on Build GRCh38
rs751218690 Chr.11: 57497803-57497803 on Build GRCh38
rs765781783 Chr.11: 57497925-57497925 on Build GRCh38
rs772283266 Chr.11: 57497760-57497760 on Build GRCh38
rs765737559 Chr.11: 57497872-57497872 on Build GRCh38
rs1046821481 Chr.11: 57497792-57497792 on Build GRCh38
rs755355570 Chr.11: 57497920-57497920 on Build GRCh38

View Full Product Details