Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183906828-183907073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STOX2
Entrez Gene ID
56977
Gene Name
storkhead box 2
Gene Aliases
-
UniGene
Hs.21958
Gene Chromosome Location
Chr.4: 183796331-184023530 on Build GRCh38
Transcripts
NM_020225.2 NR_132761.1 XM_011532129.1 XM_011532130.1 XM_017008466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1566849 c.147G>A p.S49S Chr.4: 183906937-183906937 on Build GRCh38
5776541 c.63G>T p.R21R Chr.4: 183906853-183906853 on Build GRCh38
5779097 c.104G>A p.R35H Chr.4: 183906894-183906894 on Build GRCh38
4730644 c.54_65del12 p.A22_R25delASDR Chr.4: 183906844-183906855 on Build GRCh38
1053640 c.134C>T p.A45V Chr.4: 183906924-183906924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755705509 Chr.4: 183906993-183907004 on Build GRCh38
rs922541635 Chr.4: 183906892-183906892 on Build GRCh38
rs371844183 Chr.4: 183907002-183907002 on Build GRCh38
rs771180242 Chr.4: 183906857-183906857 on Build GRCh38
rs774617929 Chr.4: 183906859-183906859 on Build GRCh38
rs764915371 Chr.4: 183906973-183906973 on Build GRCh38
rs935833213 Chr.4: 183906925-183906925 on Build GRCh38
rs1019201693 Chr.4: 183906911-183906911 on Build GRCh38
rs564124660 Chr.4: 183906998-183906998 on Build GRCh38
rs996787824 Chr.4: 183906851-183906851 on Build GRCh38
rs541264702 Chr.4: 183906987-183906987 on Build GRCh38
rs761524389 Chr.4: 183906969-183906969 on Build GRCh38
rs372362393 Chr.4: 183906852-183906852 on Build GRCh38
rs986874847 Chr.4: 183906905-183906905 on Build GRCh38
rs933949210 Chr.4: 183906907-183906907 on Build GRCh38
rs148385537 Chr.4: 183906965-183906965 on Build GRCh38
rs939113136 Chr.4: 183907025-183907025 on Build GRCh38
rs772273575 Chr.4: 183906894-183906894 on Build GRCh38
rs541537786 Chr.4: 183906916-183906916 on Build GRCh38
rs746220096 Chr.4: 183906861-183906861 on Build GRCh38
rs750236853 Chr.4: 183906983-183906983 on Build GRCh38
rs926288159 Chr.4: 183907010-183907010 on Build GRCh38
rs372942091 Chr.4: 183906963-183906963 on Build GRCh38
rs988096380 Chr.4: 183906937-183906937 on Build GRCh38
rs375362675 Chr.4: 183906938-183906938 on Build GRCh38
rs889663095 Chr.4: 183906918-183906918 on Build GRCh38
rs555013600 Chr.4: 183906928-183906928 on Build GRCh38

View Full Product Details