Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118373397-118373626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE4A
Entrez Gene ID
9354
Gene Name
ubiquitination factor E4A
Gene Aliases
E4, UBOX2, UFD2
UniGene
Hs.602967
Gene Chromosome Location
Chr.11: 118359581-118399211 on Build GRCh38
Transcripts
NM_001204077.1 NM_004788.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
28475 c.1025G>T p.S342I Chr.11: 118373573-118373573 on Build GRCh38
923553 c.1046C>A p.T349N Chr.11: 118373594-118373594 on Build GRCh38
3415570 c.1002G>T p.K334N Chr.11: 118373550-118373550 on Build GRCh38
923552 c.1019T>G p.I340S Chr.11: 118373567-118373567 on Build GRCh38
4987544 c.1028T>C p.I343T Chr.11: 118373576-118373576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771016739 Chr.11: 118373514-118373514 on Build GRCh38
rs756539733 Chr.11: 118373498-118373498 on Build GRCh38
rs754342966 Chr.11: 118373489-118373490 on Build GRCh38
rs752807213 Chr.11: 118373511-118373511 on Build GRCh38
rs370326273 Chr.11: 118373451-118373451 on Build GRCh38
rs766174530 Chr.11: 118373483-118373483 on Build GRCh38
rs757864843 Chr.11: 118373524-118373525 on Build GRCh38
rs961625382 Chr.11: 118373494-118373494 on Build GRCh38
rs764426268 Chr.11: 118373550-118373550 on Build GRCh38
rs763875115 Chr.11: 118373447-118373447 on Build GRCh38
rs201668878 Chr.11: 118373576-118373576 on Build GRCh38
rs751268784 Chr.11: 118373596-118373596 on Build GRCh38
rs375387915 Chr.11: 118373449-118373449 on Build GRCh38
rs200966232 Chr.11: 118373556-118373556 on Build GRCh38
rs777932100 Chr.11: 118373507-118373507 on Build GRCh38
rs984271272 Chr.11: 118373461-118373461 on Build GRCh38
rs369074050 Chr.11: 118373490-118373490 on Build GRCh38
rs200152975 Chr.11: 118373491-118373491 on Build GRCh38
rs776366628 Chr.11: 118373525-118373525 on Build GRCh38
rs910053436 Chr.11: 118373486-118373486 on Build GRCh38
rs891642693 Chr.11: 118373598-118373598 on Build GRCh38
rs749850965 Chr.11: 118373459-118373459 on Build GRCh38
rs748672075 Chr.11: 118373495-118373495 on Build GRCh38
rs201361239 Chr.11: 118373587-118373587 on Build GRCh38
rs749377737 Chr.11: 118373509-118373509 on Build GRCh38
rs751252185 Chr.11: 118373487-118373487 on Build GRCh38
rs142428336 Chr.11: 118373524-118373524 on Build GRCh38
rs972939996 Chr.11: 118373500-118373500 on Build GRCh38
rs760856242 Chr.11: 118373540-118373540 on Build GRCh38
rs922886158 Chr.11: 118373505-118373505 on Build GRCh38
rs757898555 Chr.11: 118373474-118373474 on Build GRCh38
rs761829625 Chr.11: 118373553-118373553 on Build GRCh38
rs773165757 Chr.11: 118373572-118373572 on Build GRCh38
rs764831511 Chr.11: 118373457-118373457 on Build GRCh38
rs374969961 Chr.11: 118373597-118373597 on Build GRCh38

View Full Product Details