Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49247243-49247516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNFX1
Entrez Gene ID
57169
Gene Name
zinc finger NFX1-type containing 1
Gene Aliases
-
UniGene
Hs.371794
Gene Chromosome Location
Chr.20: 49245897-49278244 on Build GRCh38
Transcripts
NM_021035.2 XM_006723836.1 XM_011528924.1 XM_017027971.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2761872 c.5603C>T p.T1868M Chr.20: 49247421-49247421 on Build GRCh38
3547515 c.5715C>T p.A1905A Chr.20: 49247309-49247309 on Build GRCh38
4745266 c.5752A>G p.M1918V Chr.20: 49247272-49247272 on Build GRCh38
5614787 c.5674G>A p.E1892K Chr.20: 49247350-49247350 on Build GRCh38
724272 c.5709C>A p.D1903E Chr.20: 49247315-49247315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199976903 Chr.20: 49247303-49247303 on Build GRCh38
rs769577753 Chr.20: 49247383-49247383 on Build GRCh38
rs762129198 Chr.20: 49247434-49247434 on Build GRCh38
rs746078834 Chr.20: 49247352-49247352 on Build GRCh38
rs777934331 Chr.20: 49247332-49247332 on Build GRCh38
rs767986153 Chr.20: 49247313-49247313 on Build GRCh38
rs755911230 Chr.20: 49247476-49247476 on Build GRCh38
rs774211005 Chr.20: 49247417-49247417 on Build GRCh38
rs773169695 Chr.20: 49247405-49247405 on Build GRCh38
rs760033410 Chr.20: 49247312-49247312 on Build GRCh38
rs772548118 Chr.20: 49247279-49247279 on Build GRCh38
rs1052586475 Chr.20: 49247408-49247408 on Build GRCh38
rs758565832 Chr.20: 49247471-49247471 on Build GRCh38
rs777427432 Chr.20: 49247405-49247406 on Build GRCh38
rs777626035 Chr.20: 49247483-49247483 on Build GRCh38
rs773364247 Chr.20: 49247474-49247474 on Build GRCh38
rs116689515 Chr.20: 49247420-49247420 on Build GRCh38
rs764553505 Chr.20: 49247449-49247449 on Build GRCh38
rs368406500 Chr.20: 49247344-49247344 on Build GRCh38
rs181876800 Chr.20: 49247450-49247450 on Build GRCh38
rs757387154 Chr.20: 49247451-49247451 on Build GRCh38
rs770549003 Chr.20: 49247412-49247412 on Build GRCh38
rs748137419 Chr.20: 49247381-49247381 on Build GRCh38
rs750655445 Chr.20: 49247466-49247466 on Build GRCh38
rs931641371 Chr.20: 49247300-49247300 on Build GRCh38
rs765297354 Chr.20: 49247460-49247460 on Build GRCh38
rs753826151 Chr.20: 49247333-49247333 on Build GRCh38
rs369071940 Chr.20: 49247282-49247282 on Build GRCh38
rs373022587 Chr.20: 49247342-49247342 on Build GRCh38
rs761279546 Chr.20: 49247447-49247447 on Build GRCh38
rs753254600 Chr.20: 49247314-49247314 on Build GRCh38
rs775671046 Chr.20: 49247443-49247443 on Build GRCh38
rs768507675 Chr.20: 49247368-49247368 on Build GRCh38
rs535646247 Chr.20: 49247307-49247307 on Build GRCh38
rs954911283 Chr.20: 49247277-49247277 on Build GRCh38
rs900140016 Chr.20: 49247271-49247271 on Build GRCh38
rs184446473 Chr.20: 49247415-49247415 on Build GRCh38
rs762414724 Chr.20: 49247411-49247411 on Build GRCh38
rs756523437 Chr.20: 49247323-49247323 on Build GRCh38
rs768418704 Chr.20: 49247270-49247270 on Build GRCh38
rs776609562 Chr.20: 49247275-49247275 on Build GRCh38
rs190461938 Chr.20: 49247421-49247421 on Build GRCh38
rs749241492 Chr.20: 49247492-49247492 on Build GRCh38
rs372355787 Chr.20: 49247475-49247475 on Build GRCh38
rs761586932 Chr.20: 49247276-49247276 on Build GRCh38
rs780905965 Chr.20: 49247369-49247369 on Build GRCh38
rs751229991 Chr.20: 49247311-49247311 on Build GRCh38

View Full Product Details